Skip to Content
Merck
All Photos(1)

Key Documents

D27004

Sigma-Aldrich

N,N′-Diphenylethylenediamine

98%

Synonym(s):

1,2-Dianilinoethane, N,N′-Ethylenedianiline, Wanzlick’s Reagent for aldehydes

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHCH2CH2NHC6H5
CAS Number:
Molecular Weight:
212.29
Beilstein:
646740
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

65-67 °C (lit.)

SMILES string

C(CNc1ccccc1)Nc2ccccc2

InChI

1S/C14H16N2/c1-3-7-13(8-4-1)15-11-12-16-14-9-5-2-6-10-14/h1-10,15-16H,11-12H2

InChI key

NOUUUQMKVOUUNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N′-Diphenylethylenediamine can be used:
  • To prepare nickel(II) chelates to study their chemical reactivities.
  • To prepare N-heterocyclic carbene (NHC) adducts by reacting with substituted benzaldehydes.
  • As a starting material to prepare substituted cyclic poly(methyl methacrylate)s.

Other Notes

Remainder mainly 1,4-diphenylpiperazine

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshitane Imai et al.
Chemical communications (Cambridge, England), (10)(10), 1070-1072 (2006-03-04)
By using (1R,2R)-1,2-diphenylethylenediamine as a single enantiopure compound, we achieved a novel successive optical resolution of more than one kind of racemic compound through supramolecular crystallization.
Irina L Tourkova et al.
Laboratory investigation; a journal of technical methods and pathology, 97(9), 1072-1083 (2017-07-25)
To improve definition of the physical and hormonal support of bone formation, we studied differentiation of human osteoblasts in vitro at varying combinations of ACTH, 1α,25-dihydroxyvitamin D
Synthesis of cyclic poly (methyl methacrylate) by the intramolecular cyclization of α-amino, ω-carboxyl heterodifunctional poly (methyl methacrylate)
Kubo M, et al.
Polymer Bull., 47(1), 25-30 (2001)
P J Bednarski et al.
Drug metabolism and disposition: the biological fate of chemicals, 22(3), 419-427 (1994-05-01)
The cisplatin analog [meso-1,2-bis(2,6-dichloro-4-hydroxyphenyl) ethylenediamine]dichloroplatinum(II) [PtCl2(1)], by virtue of its estrogenic 1,2-diphenylethylenediamine ligand 1, was intended to function as a cytotoxic estrogen. This article reports on the reversible and irreversible interactions of this compound with plasma and plasma proteins in
Synthesis and spectroscopic investigation of some nickel (II) chelates containing β-ketoenolates and N,N′-diphenylethylenediamine
Tsiamis C and Themeli M
Polyhedron, 13(2), 281-289 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service