Skip to Content
Merck
All Photos(1)

Key Documents

D221503

Sigma-Aldrich

2-Dodecanol

99%

Synonym(s):

Decyl methyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9CH(OH)CH3
CAS Number:
Molecular Weight:
186.33
Beilstein:
1720036
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.44 (lit.)

bp

249-250 °C (lit.)

density

0.829 g/mL at 20 °C (lit.)

SMILES string

CCCCCCCCCCC(C)O

InChI

1S/C12H26O/c1-3-4-5-6-7-8-9-10-11-12(2)13/h12-13H,3-11H2,1-2H3

InChI key

XSWSEQPWKOWORN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ankita Asati et al.
Journal of chromatography. A, 1513, 157-171 (2017-07-25)
Two low density organic solvents based liquid-liquid microextraction methods, namely Vortex assisted liquid-liquid microextraction based on solidification of floating organic droplet (VALLME-SFO) and Dispersive liquid-liquid microextraction based on solidification of floating organic droplet(DLLME-SFO) have been compared for the determination of
Mojtaba Rahimi Moghadam et al.
Analytical methods : advancing methods and applications, 12(44), 5332-5343 (2020-10-27)
This study was conducted to synthesize magnetic nanoparticles (MNPs) modified with generation 5 (G5) polyamidoamine (PAMAM) dendrimer and apply them as a sorbent in the dispersive-micro-solid phase extraction (D-μ-SPE) method for preconcentration of metribuzin in water samples. The characterization of
Kapil Mandrah et al.
Journal of chromatography. A, 1528, 10-17 (2017-11-04)
In the present study, a method has been efficiently developed for the first time to determine nine bisphenol analogues [bisphenol A (BPA), bisphenol C (BPC), bisphenol AF (BPAF), bisphenol E (BPE), bisphenol F (BPF), bisphenol G (BPG), bisphenol M (BPM)
Phitchan Sricharoen et al.
Journal of separation science, 40(19), 3848-3856 (2017-07-28)
A new approach applying a pet fish air pump is introduced to develop an extraction method, namely, air-pump-enhanced emulsion, followed by salt-assisted emulsion breaking based on solidified floating organic drop microextraction for the extraction and preconcentration of Sudan I-IV before
Rakesh Roshan Jha et al.
Journal of separation science, 40(13), 2694-2702 (2017-05-06)
Antibiotics that are used excessively and disposed of improperly are categorized as emerging pollutants. The determination of micropollutants in water with an accurate and precise method is always a big challenge. Hence, a simple, rapid, sensitive, economical and almost eco-friendly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service