Skip to Content
Merck
All Photos(5)

Key Documents

D152927

Sigma-Aldrich

Dimethyl carbonate

greener alternative

ReagentPlus®, 99%

Synonym(s):

Carbonic acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2CO
CAS Number:
Molecular Weight:
90.08
Beilstein:
635821
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Assay:
99%
bp:
90 °C (lit.)
vapor pressure:
18 mmHg ( 21.1 °C)

vapor density

3.1 (vs air)

Quality Level

vapor pressure

18 mmHg ( 21.1 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Safer Solvents and Auxiliaries
Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.368 (lit.)

bp

90 °C (lit.)

mp

2-4 °C (lit.)

density

1.069 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

O=C(OC)OC

InChI

1S/C3H6O3/c1-5-3(4)6-2/h1-2H3

InChI key

IEJIGPNLZYLLBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl carbonate (DMC) is a versatile, non-toxic, biodegradable reagent with tunable chemical reactivity. It serves as a green alternative to dimethyl sulfate or methyl halides and phosgene for methylation and carboxylation reactions. The base-catalyzed reaction between carbon dioxide and methanol to form DMC has been studied. Various thermodynamic parameters of DMC have been reported.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is a Greener alternative to conventional solvents and chemicals. Click here for more information.

Application

Dimethyl carbonate may be used to synthesize:
  • Methyl phenyl carbonate by transesterification with phenol.
  • Diphenyl carbonate by transesterification with methyl phenyl carbonate.
  • Methyl carbamates, a raw material for isocyanate synthesis.
  • Tetramethoxysilane by reacting with silica at 550-600K.

Features and Benefits

Greener solvent

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct synthesis of dimethyl carbonate from carbon dioxide and methanol catalyzed by base.
Fang S and Fujimoto K.
Applied Catalysis A: General, 142(1), L1-L3 (1996)
Milad Abolhasani et al.
Lab on a chip, 12(9), 1611-1618 (2012-03-15)
We present an automated microfluidic (MF) approach for the systematic and rapid investigation of carbon dioxide (CO(2)) mass transfer and solubility in physical solvents. Uniformly sized bubbles of CO(2) with lengths exceeding the width of the microchannel (plugs) were isothermally
Zul Ilham et al.
Bioresource technology, 101(8), 2735-2740 (2009-11-26)
This study reports on a novel two-step process for biodiesel production consisting of hydrolysis of oils in sub-critical water and subsequent supercritical dimethyl carbonate esterification. This process found to occur optimally at the sub-critical water treatment (270 degrees Celsius/27 MPa)
Jana Engeldinger et al.
Physical chemistry chemical physics : PCCP, 14(7), 2183-2191 (2011-11-18)
The simultaneous combination of steady state isotopic transient kinetic analysis (SSITKA) with diffuse reflectance Fourier transform spectroscopy (DRIFTS) and mass spectrometric (MS) analysis was applied to study the oxidative carbonylation of methanol (MeOH) to dimethyl carbonate (DMC) on a CuY
Liping Zhang et al.
Bioresource technology, 101(21), 8144-8150 (2010-07-02)
The transesterification of palm oil with dimethyl carbonate (DMC) for preparing biodiesel has been studied in solvent-free system at the catalysis of potassium hydroxide (KOH) as heterogeneous catalyst. Fatty acid methyl esters (FAMEs) were analyzed by GC with internal standard

Articles

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Related Content

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service