All Photos(2)
About This Item
Linear Formula:
(HOCH2)3CCH2OCH2C(CH2OH)3
CAS Number:
Molecular Weight:
254.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
grade
technical grade
form
powder
mp
215-218 °C (lit.)
SMILES string
OCC(CO)(CO)COCC(CO)(CO)CO
InChI
1S/C10H22O7/c11-1-9(2-12,3-13)7-17-8-10(4-14,5-15)6-16/h11-16H,1-8H2
InChI key
TXBCBTDQIULDIA-UHFFFAOYSA-N
Related Categories
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Nazmiye Dönmez et al.
Dental materials journal, 38(2), 218-225 (2018-12-07)
The micro-tensile bond strengths (micro-TBSs) of four universal adhesive systems, applied in two different bonding techniques, to carries-affected dentin after Er:YAG laser irradiation were investigated. Twenty-four extracted human molars were divided into four groups, according to the adhesive systems used.
Uzay Koc Vural et al.
Journal of prosthodontics : official journal of the American College of Prosthodontists, 28(1), e204-e209 (2017-09-30)
To investigate the etiologic factors, effect of age and gender on dental trauma, and to evaluate the performance of composite resin on the fractured crown of permanent anterior teeth. Over a 2-year period, 73 permanent teeth from 51 patients with
Isabel Cristina Olegário et al.
Clinical oral investigations, 23(4), 1761-1770 (2018-09-02)
To evaluate the survival of atraumatic restorative treatment (ART) restorations using high viscosity glass ionomer cement (GIC), compomer (COM), and glass carbomer (CAR) for occlusal and occlusoproximal cavitated dentin caries lesions in primary molars. A total of 568 4-7-year-old children
Satoshi Fujiwara et al.
Journal of the mechanical behavior of biomedical materials, 77, 501-509 (2017-10-19)
The aim of this study was to determine the effect of double-layer application of universal adhesives on the bond quality and compare to other adhesive systems. Two universal adhesives used were in this study: Scotchbond Universal (SU), [3M ESPE] and
Mario F Gutiérrez et al.
Journal of dentistry, 82, 45-55 (2019-02-11)
To evaluate the effect of addition of zinc oxide and copper nanoparticles (ZnO/CuNp) into universal adhesives, on antimicrobial activity (AMA), cytotoxicity (CTX), water sorption (WS) and solubility (SO), microhardness (MH) and in vitro degree of conversion (DC), as well as
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service