P4755
Pentaerythritol
98%
Synonym(s):
2,2-Bis(hydroxymethyl)-1,3-propanediol
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
C(CH2OH)4
CAS Number:
Molecular Weight:
136.15
Beilstein:
1679274
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor pressure
<1 mmHg ( 20 °C)
Quality Level
Assay
98%
form
powder
bp
276 °C/30 mmHg (lit.)
mp
253-258 °C (lit.)
SMILES string
OCC(CO)(CO)CO
InChI
1S/C5H12O4/c6-1-5(2-7,3-8)4-9/h6-9H,1-4H2
InChI key
WXZMFSXDPGVJKK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Pentaerythritol is a versatile starting material to synthesize various dendrimers and star-shaped polymers. It is widely used in the preparation of flame-retardant epoxy resins and polymer composites. It is also used in the synthesis of the vasodilator, pentaerythritol tetranitrate (PETN).
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A novel intumescent flame-retardant polyethylene system.
Xie, Fei et al.
Macromolecular Materials and Engineering, 291(3), 247-253 (2006)
Supramolecular self-organization of ?Janus-like? diblock codendrimers: synthesis, thermal behavior, and phase structure modeling.
Bury, Izabela et al.
Chemistry?A European Journal , 12(32), 8396-8413 (2006)
NO donors. Part 18: Bioactive metabolites of GTN and PETN?Synthesis and vasorelaxant properties.
Lange, Kathrin et al.
Bioorganic & Medicinal Chemistry, 19(11), 3141-3144 (2009)
Flame retardancy and thermal degradation mechanism of epoxy resin composites based on a DOPO substituted organophosphorus oligomer.
Wang, Xin et al.
Polymer, 51(11), 2435-2445 (2010)
Thomas Vanbergen et al.
ChemSusChem (2020-05-30)
Split-phase alcoholysis of flexible polyurethane (PU) foam yields an apolar phase containing the recycled polyether polyol, and a lower, polar phase of the alcoholyzing agent and aromatic compounds. However, multiple purification steps are required to render the polyether polyol suitable
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service