253847
2,3,4-Trihydroxybenzoic acid
97%
Synonym(s):
Pyrogallol-4-carboxylic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(HO)3C6H2CO2H
CAS Number:
Molecular Weight:
170.12
Beilstein:
2210807
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
mp
~205 °C (dec.) (lit.)
functional group
carboxylic acid
SMILES string
OC(=O)c1ccc(O)c(O)c1O
InChI
1S/C7H6O5/c8-4-2-1-3(7(11)12)5(9)6(4)10/h1-2,8-10H,(H,11,12)
InChI key
BRRSNXCXLSVPFC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
DPPH radical-scavenging ability of 2,3,4-trihydroxybenzoic acid has been evaluated.
Application
2,3,4-Trihydroxybenzoic acid has been used as:
- reagent during catalytic spectrophotometric determination of vanadium in natural and sea waters
- internal standard in separation of phenolic acids by HPLC with photodiode array detection
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Rebecca J Robbins et al.
Journal of chromatography. A, 1038(1-2), 97-105 (2004-07-06)
A quantitative high-performance liquid chromatography-photodiode array detection method separating 16 phenolic acids was achieved. Six columns and several mobile phases were investigated. Resolution was achieved with a high-purity silica Phenomenex Luna C18 column (150 mm x 4.6 mm, 5 microm)
F Ramezani Ali Akbari et al.
Acta endocrinologica (Bucharest, Romania : 2005), 15(2), 187-194 (2019-09-12)
Endothelial dysfunction and diabetic cardiomyopathy are critical complications of diabetes. Gallic acid (GA) plays a significant role in cardiovascular disorders resulted from diabetes. In addition, increased plasma miR-24, miR-126 associated with endothelial dysfunction. The current study was designed to assess
Yuan Gao et al.
Journal of the science of food and agriculture, 99(7), 3555-3562 (2019-01-11)
Sweet cherries (Prunus avium L.) are rich in polyphenols and have high antioxidant potential. This study deals with the use of ultra-performance liquid chromatography-tandem mass spectrometry on five cherry cultivars, namely Caihong, Sunburst, Summit, 23-51 and Valeri, to determine the
Ana Maria Buljat et al.
Food technology and biotechnology, 57(2), 159-170 (2019-09-21)
The foam mat drying process is a simple and economical alternative to spray and freeze drying. It is considerably cheaper due to rapid drying at low temperatures, preservation of nutritional quality and easy reconstitution. Basic principle of the process is
Yaghoob Farbood et al.
Avicenna journal of phytomedicine, 3(4), 329-340 (2014-07-23)
It is generally agreed that most of the phenomena observed during brain ischemia and reperfusion can be explained by the damage to membrane structure. Oxidative stress is resulted in an imbalance between high consumption of oxygen and low levels of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service