Skip to Content
Merck
All Photos(1)

Key Documents

CDS010546

Sigma-Aldrich

phenylphosphoric acid

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7O4P
Molecular Weight:
174.09
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

form

solid

SMILES string

OP(O)(=O)Oc1ccccc1

InChI

1S/C6H7O4P/c7-11(8,9)10-6-4-2-1-3-5-6/h1-5H,(H2,7,8,9)

InChI key

CMPQUABWPXYYSH-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen Klap et al.
Plants (Basel, Switzerland), 9(5) (2020-05-20)
During 2019, tomato fruits showing viral-like symptoms of marbled yellow spots were abundant in Israel. The new symptoms were distinctive from those typical of tomato brown rugose fruit virus (ToBRFV) infection but resembled symptoms of pepino mosaic virus (PepMV) infection.
Kentaro Miyazaki
PloS one, 13(4), e0194090-e0194090 (2018-04-12)
I have previously identified a metagenomic fragment (~4 kb) containing the salicylate (2-hydroxybenzoate)-responsive transcriptional regulator Sal7AR. Taking advantage of the inert nature of salicylate to common genetic switches used in Escherichia coli, here I developed a salicylate-inducible high expression system
J M Ashraf et al.
Biochemistry. Biokhimiia, 82(5), 579-586 (2017-06-12)
Advanced glycation end-products (AGEs) have been found to be critically involved in initiation or progression of diabetes secondary complications (nephropathy, retinopathy, neuropathy, and angiopathy). Various hyper-glycating carbonyl compounds such as 3-deoxyglucosone (3-DG) are produced in pathophysiological conditions that form AGEs
J K Akintunde et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 126, 110043-110043 (2020-03-17)
People who have experienced high blood pressure are at greater risk of susceptibility to other health problems including oculopathy. The patients with these experiences do not have adequate treatment and those who do; spend much funds on the drug purchase.
Junyi Zhao et al.
Planta, 248(4), 1017-1027 (2018-07-22)
ALDH7B4 expression contributes to abiotic stress tolerance. The NAC transcription factor ATAF1 is a main regulator of expression of the ALDH7B4 gene in Arabidopsis thaliana as shown by ATAF1 mutants. The aldehyde dehydrogenase 7B4 (ALDH7B4) protein has important roles in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service