Skip to Content
Merck
All Photos(1)

Documents

CDS000560

Sigma-Aldrich

Phenyl propionate

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O2
CAS Number:
Molecular Weight:
150.17
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

liquid

SMILES string

CCC(=O)Oc1ccccc1

InChI

1S/C9H10O2/c1-2-9(10)11-8-6-4-3-5-7-8/h3-7H,2H2,1H3

InChI key

DYUMLJSJISTVPV-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kamil Stastny et al.
Metabolites, 10(8) (2020-07-31)
The use of anabolic steroid hormones as growth promoters in feed for farm animals has been banned in the European Union since 1988 on the basis of Council Directive 96/22/EC. However, there is still ongoing monitoring and reporting of positive
Sven C Mueller et al.
The journal of sexual medicine, 17(12), 2495-2507 (2020-11-24)
Whereas mounting work has begun to document the neural correlates underlying sexual arousal (SA) in humans, the associations between gender identity and the brain correlates of SA as well as their hormonal contributions remain unknown. This study investigated neural activation
Eva Maria Prem et al.
Biotechnology for biofuels, 14(1), 27-27 (2021-01-22)
Lignin intermediates resulting from lignocellulose degradation have been suspected to hinder anaerobic mineralisation of organic materials to biogas. Phenyl acids like phenylacetate (PAA) are early detectable intermediates during anaerobic digestion (AD) of aromatic compounds. Studying the phenyl acid formation dynamics
Eva Maria Prem et al.
Biotechnology for biofuels, 13, 81-81 (2020-05-12)
Proteinaceous wastes exhibit high theoretical methane yields and their residues are considered valuable fertilisers. The routine anaerobic degradation of proteins often raises problems like high aromatic compound concentrations caused by the entry of aromatic amino acids into the system. A
Nicolas Fabresse et al.
International journal of legal medicine, 131(4), 989-999 (2017-02-27)
We developed and validated a method to detect and quantify 12 anabolic steroids in blood (androstenedione, dihydrotestosterone, boldenone, epitestosterone, mesterolone, methandienone, nandrolone, stanozolol, norandrostenedione, tamoxifene, testosterone, trenbolone) and eight more in hair samples (nandrolone phenylpropionate, nandrolone decanoate, testosterone propionate, testosterone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service