Skip to Content
Merck
All Photos(1)

Documents

108723

Sigma-Aldrich

Phenyl acetate

99%

Synonym(s):

Acetic acid phenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC6H5
CAS Number:
Molecular Weight:
136.15
Beilstein:
636458
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.501 (lit.)

bp

196 °C (lit.)

density

1.073 g/mL at 25 °C (lit.)

SMILES string

CC(=O)Oc1ccccc1

InChI

1S/C8H8O2/c1-7(9)10-8-5-3-2-4-6-8/h2-6H,1H3

InChI key

IPBVNPXQWQGGJP-UHFFFAOYSA-N

Gene Information

human ... PON1(5444)

Looking for similar products? Visit Product Comparison Guide

General description

Phenyl acetate is an aromatic ester. Phenyl acetate levels in urine are marker for the diagnosis of some forms of unipolar major depressive disorders. It undergoes Fries rearrangement to form a mixture of o- and p-hydroxyacetophenones which are useful intermediates in manufacture of pharmaceuticals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristiano Noto et al.
Journal of psychiatric research, 68, 210-216 (2015-08-01)
Schizophrenia is accompanied by increased lipid peroxidation and nitric oxide (NO) levels and by lowered antioxidant levels. However, the effect of antipsychotic agents on these processes remains unclear. The objective of this study is to determine the oxidative stress (OS)
Vasilios Hatzistavros et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 21(7), 823-826 (2005-07-26)
Selective membranes containing a few micrograms of various complexing reagents in a polyvinyl chloride (PVC) matrix were produced in order to determine low uranium concentrations in water. The membranes were produced on the surfaces of quartz glasses, and immersed in
Ferhat Içme et al.
Turkish journal of medical sciences, 45(4), 947-953 (2015-10-02)
The aims of this study were to investigate the significance of oxidative stress parameters in the pathogenesis of ischemic stroke and hemorrhagic stroke and to investigate their effects on stroke severity using the National Institutes of Health Stroke Scale (NIHSS).
Solvent effects in liquid phase Fries rearrangement of phenyl acetate over a HBEA zeolite.
Jayat F, et al.
Catalysis Letters, 41(3-4), 181-187 (1996)
A Gugliucci et al.
Clinica chimica acta; international journal of clinical chemistry, 445, 41-47 (2015-03-25)
Recent studies showing that high density lipoproteins (HDL) can effect plaque regression indicate that recent trial failures do not exclude an atheroprotective role of HDL. Instead, they highlight differences between HDL function and measured HDL-cholesterol (HDL-C). PON1 is one key

Articles

Quantum dots (QDs): Semiconductor nanoparticles with diverse applications including displays, lighting, and biomedical imaging.

Quantum dots (QDs): Semiconductor nanoparticles with diverse applications including displays, lighting, and biomedical imaging.

Quantum dots (QDs): Semiconductor nanoparticles with diverse applications including displays, lighting, and biomedical imaging.

Quantum dots (QDs): Semiconductor nanoparticles with diverse applications including displays, lighting, and biomedical imaging.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service