Skip to Content
Merck
All Photos(3)

Key Documents

C121800

Sigma-Aldrich

L-Cysteine hydrochloride hydrate

99%, for peptide synthesis

Synonym(s):

L-Cysteine·HCl hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(NH2)CO2H·HCl·xH2O
CAS Number:
Molecular Weight:
157.62 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Product Name

L-Cysteine hydrochloride hydrate, 99%

Assay

99%

form

solid

optical activity

[α]23/D +5.2°, c = 2 in 5 M HCl

reaction suitability

reaction type: solution phase peptide synthesis

technique(s)

affinity chromatography: suitable

color

white

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

Cl[H].[H]O[H].N[C@@H](CS)C(O)=O

InChI

1S/C3H7NO2S.ClH.H2O/c4-2(1-7)3(5)6;;/h2,7H,1,4H2,(H,5,6);1H;1H2/t2-;;/m0../s1

InChI key

QIJRTFXNRTXDIP-JIZZDEOASA-N

General description

L-Cysteine hydrochloride hydrate also known as L-Cysteine·HCl hydrate, is commonly used as a starting material in the synthesis of L-cysteine (alkylated, acidic, and alcoholic) derivatives. [1]

Application

L-Cysteine hydrochloride hydrate is used as a standard solution to determine the quantity of thiol groups attached on mucoadhesiveness. [2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cysteine based novel noncompetitive inhibitors of urease (s)?Distinctive inhibition susceptibility of microbial and plant ureases
Z Amtul
Bioorganic & Medicinal Chemistry, 14, 6737-6744 (2006)
Mucoadhesive thiolated chitosans as platforms for oral controlled drug delivery: synthesis and in vitro evaluation
M Roldo
European Journal of Pharmaceutics and Biopharmaceutics, 57, 115-121 (2004)
Ana Odetth Quintana-Escobar et al.
Plants (Basel, Switzerland), 12(24) (2023-12-23)
Plant growth regulators (PGR) are essential for somatic embryogenesis (SE) in different species, and Coffea canephora is no exception. In our study model, previously, we have been able to elucidate the participation of various genes involved in SE by using
Shokouh Ahmadi et al.
Journal of visualized experiments : JoVE, (149) (2019-08-20)
The emerging role of the gut microbiome in several human diseases demands a breakthrough of new tools, techniques and technologies. Such improvements are needed to decipher the utilization of microbiome modulators for human health benefits. However, the large-scale screening and
Gerdt Müller et al.
Biotechnology for biofuels, 8, 187-187 (2015-11-27)
The emerging bioeconomy depends on improved methods for processing of lignocellulosic biomass to fuels and chemicals. Saccharification of lignocellulose to fermentable sugars is a key step in this regard where enzymatic catalysis plays an important role and is a major

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service