B52808
1,2-Di(4-pyridyl)ethylene
97%
Synonym(s):
1,2-Bis(4-pyridyl)ethylene, 4,4′-Vinylenedipyridine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H10N2
CAS Number:
Molecular Weight:
182.22
Beilstein:
3865
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
crystals
mp
148-152 °C (lit.)
SMILES string
c1cc(ccn1)\C=C\c2ccncc2
InChI
1S/C12H10N2/c1(11-3-7-13-8-4-11)2-12-5-9-14-10-6-12/h1-10H/b2-1+
InChI key
MGFJDEHFNMWYBD-OWOJBTEDSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
1,2-Di(4-pyridyl)ethylene is extensively used as a ligand/linker in the synthesis of a variety of metal-organic frameworks(MOFs) and coordination polymers (CPs). It is employed in the synthesis of:
- Expanding and shrinking cadmium-based pillared layer coordination polymers showing selective guest adsorption.
- Dinuclear zinc complex that exhibits photocontrolled fluorescence.
- Platinum-based 3-D tetragonal prisms via multicomponent coordination-driven self-assembly.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Zhiwei Chen et al.
Dalton transactions (Cambridge, England : 2003), 47(17), 6240-6249 (2018-04-25)
Three new luminescent transition coordination polymers, namely, {[Cd(L)(4,4-bpy)]·DMF·H2O}n (1), {[Cd2(L)2(bpe)2]·3DMF·2.5H2O}n (2), and {[Cd(L)(bibp)]·2DMF}n (3), (H2L = 4,4'-[benzene-1,3-diylbis(methanediylsulfanediyl)]dibenzoic acid, 4,4-bpy = 4,4-bipyridine, bpe = 1,2-bis(4-pyridyl)ethylene, and bibp = 4,4'-bis(benzoimidazo-1-ly)biphenyl), were solvothermally synthesized using Cd2+ ions and S-containing dicarboxylate acid in the
Isoreticular Homochiral Porous Metal- Organic Structures with Tunable Pore Sizes.
Dybtsev DN, et al.
Inorganic Chemistry, 46(17), 6843-6845 (2007)
Expanding and Shrinking Porous Modulation Based on Pillared-Layer Coordination Polymers Showing Selective Guest Adsorption.
Maji TK, et al.
Angewandte Chemie (International Edition in English), 43(25), 3269-3272 (2004)
Jun-Hui Zhang et al.
Electrophoresis, 38(19), 2513-2520 (2017-07-06)
Natural amino acids are well known to form coordination polymers with transition metal ions. In this study, six homochiral metal-organic frameworks constructed from Zn
Ekaterina Babich et al.
Nanomaterials (Basel, Switzerland), 10(9) (2020-09-20)
The irradiation of silver-to-sodium ion-exchanged glass with 1.06-μm nanosecond laser pulses of mJ-range energy results in the formation of silver nanoparticles under the glass surface. Following chemical removal of ~25-nm glass layer reveals a pattern of nanoparticles capable of surface
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service