Skip to Content
Merck
All Photos(1)

Key Documents

A76982

Sigma-Aldrich

3-Amino-2-pyrazinecarboxylic acid

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N3O2
CAS Number:
Molecular Weight:
139.11
Beilstein:
124835
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
powder
Assay:
≥99%

Quality Level

Assay

≥99%

form

powder

mp

205-210 °C (dec.) (lit.)

SMILES string

Nc1nccnc1C(O)=O

InChI

1S/C5H5N3O2/c6-4-3(5(9)10)7-1-2-8-4/h1-2H,(H2,6,8)(H,9,10)

InChI key

ZAGZIOYVEIDDJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A J Dobson et al.
Acta crystallographica. Section C, Crystal structure communications, 52 ( Pt 6), 1512-1514 (1996-06-15)
3-Aminopyrazine-2-carboxylic acid, C5H5N3O2, displays an extensive network of intra- and intermolecular hydrogen bonds which are undoubtedly responsible for the modest values of the displacement parameters. H-atom transfer to the ring N atoms did not occur and the carboxy and amino
Ghada Bouz et al.
Molecules (Basel, Switzerland), 24(7) (2019-03-31)
We report the design, synthesis, and in vitro antimicrobial activity of a series of N-substituted 3-aminopyrazine-2-carboxamides with free amino groups in position 3 on the pyrazine ring. Based on various substituents on the carboxamidic moiety, the series is subdivided into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service