Skip to Content
Merck
All Photos(3)

Key Documents

158631

Sigma-Aldrich

4′-Chloroacetanilide

97%

Synonym(s):

N-(4-Chlorophenyl)acetamide, Acetic acid 4-chloroanilide, NSC 40563, NSC 444

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHC6H4Cl
CAS Number:
Molecular Weight:
169.61
Beilstein:
509638
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

176-178 °C (lit.)

functional group

amide
chloro

SMILES string

CC(=O)Nc1ccc(Cl)cc1

InChI

1S/C8H8ClNO/c1-6(11)10-8-4-2-7(9)3-5-8/h2-5H,1H3,(H,10,11)

InChI key

GGUOCFNAWIODMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4′-Chloroacetanilide is the main impurity present in acetaminophen and has been quantitated by new high-performance liquid chromatography method using a mixed-mode reversed-phase/cation exchange stationary phase.

Application

<ul>
<li><strong>High-performance liquid chromatography (HPLC) method validation:</strong> 4 chloroacetanilide is used as an internal standard for isosorbide dinitrate in the assay of sustained-release tablets or capsules containing nitroglycerin, isosorbide dinitrate, or pentaerythritol tetranitrate by high-performance liquid chromatography (Gelber and Papas, 1983).</li>
</ul>

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

L Koymans et al.
Xenobiotica; the fate of foreign compounds in biological systems, 23(6), 633-648 (1993-06-01)
1. The general mechanism of metabolic oxidation of substrates by cytochromes P450 (P450s) appears to consist of sequential one-electron oxidation steps rather than of a single concerted transfer of activated oxygen species from P450 to substrates. 2. In case of
Tamas A Godany et al.
Chimia, 65(4), 253-255 (2011-06-18)
Lithiation of N-(4-chlorophenyl)-pivalamide (NCP) and two additional substituted acetanilides: 4-fluoroacetanilide (4-F) and 4-chloroacetanilide (4-Cl) has been monitored by means of calorimetry, on-line ATR-IR and UV/vis spectroscopy and endoscopy. The combined on-line monitoring revealed the differences between the reaction paths of
Anthony F Pizon et al.
Clinical toxicology (Philadelphia, Pa.), 47(2), 132-136 (2008-07-09)
p-Chloroaniline is more potent at producing methemoglobin than aniline in animal models. This case highlights the clinical presentation of an inhalation exposure to p-chloroaniline and associated laboratory analysis. An in-vitro study evaluating the metabolism of p-chloroaniline in human hepatocytes was
Octavian Călinescu et al.
Journal of chromatographic science, 50(4), 335-342 (2012-03-13)
Determination of acetaminophen and its main impurities: 4-nitrophenol, 4'-chloroacetanilide, as well as 4-aminophenol and its degradation products, p-benzoquinone and hydroquinone has been developed and validated by a new high-performance liquid chromatography method. Chromatographic separation has been obtained on a Hypersil
Jody A Shoemaker et al.
Journal of AOAC International, 89(1), 201-209 (2006-03-04)
U.S. Environmental Protection Agency (EPA) Method 535 has been developed in order to provide a method for the analysis of "Alachlor ESA and other acetanilide degradation products," which are listed on EPA's 1998 Drinking Water Contaminant Candidate List. Method 535

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service