Skip to Content
Merck
All Photos(1)

Key Documents

A34201

Sigma-Aldrich

Allyl methyl sulfide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2SCH3
CAS Number:
Molecular Weight:
88.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.4714 (lit.)

bp

91-93 °C (lit.)

density

0.803 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CSCC=C

InChI

1S/C4H8S/c1-3-4-5-2/h3H,1,4H2,2H3

InChI key

NVLPQIPTCCLBEU-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

64.4 °F - closed cup

Flash Point(C)

18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jen-Chieh Tsai et al.
Molecules (Basel, Switzerland), 24(6) (2019-03-23)
The mechanism of hepatoprotective compounds is usually related to its antioxidant or anti-inflammatory effects. Black garlic is produced from garlic by heat treatment and its anti-inflammatory activity has been previously reported. Therefore, the aim of this study was to investigate
Larry D Lawson et al.
Journal of agricultural and food chemistry, 53(16), 6254-6261 (2005-08-04)
In support of a new clinical trial designed to compare the effects of crushed fresh garlic and two types of garlic supplement tablets (enteric-coated dried fresh garlic and dried aged garlic extract) on serum lipids, the three garlic products have
Larry D Lawson et al.
Journal of agricultural and food chemistry, 53(6), 1974-1983 (2005-03-17)
Progress in establishing systemic pharmacological effects for fresh, crushed garlic (Allium sativum L) in humans has been hindered by (1) the inability to measure allicin bioavailability, (2) lack of direct evidence that allicin has significant systemic activity at doses of
Hsiao-Pei Chang et al.
Nutrition (Burbank, Los Angeles County, Calif.), 21(4), 530-536 (2005-04-07)
We investigated the inhibition of nitric oxide (NO) and prostaglandin E2 (PGE2) production by the garlic oil derivatives, diallyl sulfide (DAS), diallyl disulfide (DADS), and allyl methyl sulfide (AMS), in lipopolysaccharide (LPS)-activated RAW 264.7 cells. Cells were treated with LPS
Claudia Castro et al.
Molecular nutrition & food research, 54(6), 781-787 (2009-11-12)
Garlic has been used for prevention and treatment of hypertension; however, the molecular mechanisms of garlic's effects remain to be elucidated. In this study, the mechanisms of the in vitro effect of organosulphur compounds derived from garlic on growth and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service