Skip to Content
Merck
All Photos(1)

Documents

W204218

Sigma-Aldrich

Allyl sulfide

≥97%, FG

Synonym(s):

Thioallyl ether, 3,3′-Thiobis-1-propene, Diallyl sulfide, NSC 20947

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2=CHCH2)2S
CAS Number:
Molecular Weight:
114.21
FEMA Number:
2042
Beilstein:
1736016
EC Number:
Council of Europe no.:
2174
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.088
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

3.9 (vs air)

vapor pressure

7 mmHg ( 20 °C)

Assay

≥97%

refractive index

n20/D 1.490 (lit.)

bp

138 °C (lit.)

mp

−83 °C (lit.)

density

0.887 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

garlic; horseradish; onion; sulfurous

SMILES string

C=CCSCC=C

InChI

1S/C6H10S/c1-3-5-7-6-4-2/h3-4H,1-2,5-6H2

InChI key

UBJVUCKUDDKUJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Allyl sulfide is a volatile flavor compound naturally found in Allium species such as garlic and onion. It is has a characteristic garlic odor and is used as a flavoring agent in meat and condiments.

Application


  • Diallylsulfide attenuates excessive collagen production and apoptosis in a rat model of bleomycin induced pulmonary fibrosis through the involvement of protease activated receptor-2.: The research highlights the protective effects of diallyl sulfide against pulmonary fibrosis, focusing on its ability to modulate protease activated receptor-2 and reduce collagen production (Kalayarasan et al., 2013).

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burdock GA
Encyclopedia of Food and Color Additives, 1, 101-101 (1997)
Fenaroli's Handbook of Flavor Ingredients, Fifth Edition, 1971, 25-25 null
Xiaonan Lu et al.
The Journal of antimicrobial chemotherapy, 67(8), 1915-1926 (2012-05-03)
Bacterial biofilms pose significant food safety risks because of their attachment to fomites and food surfaces, including fresh produce surfaces. The purpose of this study was to systematically investigate the activity of selected antimicrobials on Campylobacter jejuni biofilms. C. jejuni
Mohamed A A Abd El-Galil et al.
Veterinary parasitology, 185(2-4), 57-63 (2011-12-06)
The present work was designed to study the prevalence of trichodinosis and gyrodactylosis in Oreochromis niloticus fries, and to test the therapeutic efficacy and preventive efficacy of garlic oil and crushed garlic cloves. Trichodinosis and gyrodactylosis are ectoparasitic diseases that
Zhi Zhu et al.
Journal of the science of food and agriculture, 93(3), 560-567 (2012-08-14)
Garlic oil (GO) has blood lipid-lowering effects. Milk fatty acid (FA) originates partly from plasma, and can be affected by the mammary lipogenesis. This study aimed to investigate GO effects on milk FA profile and mammary lipogenesis-related gene expression. Early-lactation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service