Skip to Content
Merck
All Photos(2)

Key Documents

932302

Sigma-Aldrich

Poly[(9,9-dioctylfluorenyl-2,7-diyl)-alt-(2,6-pyridine)]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C34H43N)n
CAS Number:
MDL number:
UNSPSC Code:
12352207
NACRES:
NA.23

mol wt

Mw ≥20,000 by GPC

Quality Level

solubility

THF: soluble
chlorobenzene: soluble
chloroform: soluble
dichlorobenzene: soluble

λmax

364 nm±10 nm in THF

fluorescence

λem 406 nm±20 nm in THF

Application

Polymer, Emitting Layers for Solution-Processed OLEDs; blue emission.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service