Skip to Content
Merck
All Photos(1)

Key Documents

933619

Sigma-Aldrich

(S)-N-(1-Phenyl-3-(piperidin-1-yl)propan-2-yl)acetamide

≥95%

Synonym(s):

N-[(1S)-1-(Phenylmethyl)-2-(1-piperidinyl)ethyl]acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24N2O
CAS Number:
Molecular Weight:
260.37
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21
form:
powder
Assay:
≥95%

Quality Level

Assay

≥95%

form

powder

functional group

amide
phenyl

storage temp.

−20°C

Application

(S)-N-(3-Methyl-1-(phenylthio)butan-2-yl)acetamide is a ligand developed by Jin-Quan Yu, active in Pd-based enantioselective C-H(sp3) arylation reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Probing Catalyst Speciation in Pd-MPAAM-Catalyzed Enantioselective C(sp3)?H Arylation: Catalyst Improvement via Destabilization of Off-Cycle Species
Hao, et al.
ACS Catalysis, 11, 11040?11048-11040?11048 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service