Skip to Content
Merck
All Photos(2)

Key Documents

926833

Sigma-Aldrich

1,4-Bis[4-(11-acryloyloxyundecyloxy)benzoyloxy]-2-methylbenzene

greener alternative
Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H64O10
CAS Number:
Molecular Weight:
813.03
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

description

Mesophase behaviour: Cr 72-75 SmX 86 SmY 93 I

Quality Level

form

solid

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

white to off-white

greener alternative category

storage temp.

2-8°C

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to enabling category of greener alternatives and has been enhanced for energy efficiency. 4D printing basically produces zero waste. It holds the promise in the fight against climate change and manufacturing waste. Click here  for more information.

Application

Our mesogenic diacrylatemonomer will enable development of stimuli-responsive engineered liquid crystal elastomers that can actuate with photo-thermal , thermal or pure photo-effect stimulus.
This product can be used in Material Jetting or Vat polymerization 3D printing techniques.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

All-Optical Control of Shape.
B. Donovan et al.,
Advanced Materials, 31, 1805750-1805750 (2019)
Self-sustained actuation from heat dissipation in liquid crystal polymer networks
G. Vantomme et al.
Journal of Polymer Science Part A: Polymer Chemistry, 56, 1331-1336 (2018)
Deformation and Elastic Recovery of Acrylate-Based Liquid Crystalline Elastomers.
Brannum M T, et al.
Macromolecules, 52, 8248-8255 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service