Skip to Content
Merck
All Photos(1)

Key Documents

912395

Sigma-Aldrich

NaBHT

Synonym(s):

Sodium butylated hydroxytoluene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23NaO
CAS Number:
Molecular Weight:
242.33
MDL number:
UNSPSC Code:
12352111
NACRES:
NA.22

form

powder

Quality Level

storage temp.

−20°C

SMILES string

[Na+].[O-]c1c(cc(cc1C(C)(C)C)C)C(C)(C)C

InChI

1S/C15H24O.Na/c1-10-8-11(14(2,3)4)13(16)12(9-10)15(5,6)7;/h8-9,16H,1-7H3;/q;+1/p-1

InChI key

AOIQNBCUUALSSZ-UHFFFAOYSA-M

Application

NaBHT is an organic soluble base used for the Pd-catalyzed arylation of base-sensitve amines with aryl halides. NaBHT has also been show to be an efficient hydride source for the Pd-catalyzed reduction of aryl halides.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abir Khadra et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(13), 3206-3212 (2017-01-10)
The Pd-PEPPSI-IPentCl precatalyst (PEPPSI=pyridine-enhanced precatalyst preparation stabilisation initiation) has been demonstrated to be highly effective in the coupling of hetero(aryl)chlorides to free 2-aminopyridine substrates to produce N-(hetero)aryl-2-aminopyridine derivatives. The catalyst has proven to be competent in a number of other
Sepideh Sharif et al.
Angewandte Chemie (International ed. in English), 54(33), 9507-9511 (2015-06-23)
A single set of reaction conditions for the palladium-catalyzed amination of a wide variety of (hetero)aryl halides using primary alkyl amines has been developed. By combining the exceptionally high reactivity of the Pd-PEPPSI-IPent(Cl) catalyst (PEPPSI=pyridine enhanced precatalyst preparation, stabilization, and
A General Protocol for the Broad-Spectrum Cross-Coupling of Nonactivated Sterically Hindered 1° and 2° Amines
Khadra A, et al.
Organometallics, 36 (18), 3573-3577 (2017)
Sepideh Sharif et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(57), 13099-13103 (2019-09-21)
NaBHT (sodium butylated hydroxytoluene), a hindered and soluble base for the efficient arylation of various base-sensitive amines and (hetero)aryl halides has been found to have an unanticipated role as a hydride donor to reduce (hetero)aryl halides and allylic acetates. Mechanistic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service