Skip to Content
Merck
All Photos(4)

Key Documents

732117

Sigma-Aldrich

Pd-PEPPSI-IPent catalyst

≥95%

Synonym(s):

Dichloro[1,3-bis(2,6-Di-3-pentylphenyl)imidazol-2-ylidene](3-chloropyridyl)palladium(II), [1,3-Bis(2,6-Di-3-pentylphenyl)imidazol-2-ylidene](3-chloropyridyl)dichloropalladium(II), [1,3-Bis(2,6-Di-3-pentylphenyl)imidazol-2-ylidene](3-chloropyridyl)palladium(II) dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H56Cl3N3Pd
CAS Number:
Molecular Weight:
791.67
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

195-201 °C

storage temp.

−20°C

SMILES string

Clc1cccnc1.CCC(CC)c2cccc(C(CC)CC)c2N3C=CN(c4c(cccc4C(CC)CC)C(CC)CC)\C3=[Pd](/Cl)Cl

InChI

1S/C35H52N2.C5H4ClN.2ClH.Pd/c1-9-26(10-2)30-19-17-20-31(27(11-3)12-4)34(30)36-23-24-37(25-36)35-32(28(13-5)14-6)21-18-22-33(35)29(15-7)16-8;6-5-2-1-3-7-4-5;;;/h17-24,26-29H,9-16H2,1-8H3;1-4H;2*1H;/q;;;;+2/p-2

InChI key

BCXSKTXOKALLAZ-UHFFFAOYSA-L

General description

Pd-PEPPSI-IPent catalyst is a palladium-based catalyst used in cross-coupling reactions to form C-N and C- C bonds.

Application

  • Catalyst for Stille coupling reaction (eq. 1)
  • Catalyst for Negishi coupling reaction (eq. 2)
  • Catalyst for Suzuki coupling reaction (eq. 3)
Reaction scheme for Pd catalyst PEPPSI<SUP>™</SUP>-IPent used in cross coupling reaction
For small scale and high throughput uses, product is also available as ChemBeads (928399)

Legal Information

Patented, U.S. Pat. No. 7,250,510. Sold under an exclusive license from Total Synthesis Ltd.
PEPPSI is a trademark of Total Synthesis Ltd.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael G Organ et al.
Angewandte Chemie (International ed. in English), 48(13), 2383-2387 (2009-02-20)
Incredible Bulk: A series of N-heterocyclic carbene catalysts (see picture) were prepared and evaluated in the Suzuki-Miyaura reaction. A variety of sterically encumbered tetra-ortho-substituted biaryl products were formed from unreactive aryl chlorides using the isopentyl-substituted catalyst at temperatures ranging from
Pd-PEPPSI-IPent: low-temperature negishi cross-coupling for the preparation of highly functionalized, tetra-ortho-substituted biaryls.
Selçuk Calimsiz et al.
Angewandte Chemie (International ed. in English), 49(11), 2014-2017 (2010-02-18)
Meenakshi Dowlut et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(14), 4279-4283 (2010-03-09)
The reactivity of Pd-PEPPSI (Pyridine, Enhanced, Precatalyst, Preparation, Stabilization, and Initiation) precatalysts in the Stille-Migita cross-coupling reaction between heteroaryl stannanes and aryl or heteroaryl halides was evaluated. In general, Pd-PEPPSI-IPent (IPent=diisopentylphenylimidazolium derivative) demonstrated high efficiency over a variety of challenging
Cory Valente et al.
Angewandte Chemie (International ed. in English), 51(14), 3314-3332 (2012-01-31)
Palladium-catalyzed cross-coupling reactions enable organic chemists to form C-C bonds in targeted positions and under mild conditions. Although phosphine ligands have been intensively researched, in the search for even better cross-coupling catalysts attention has recently turned to the use of
Pd-PEPPSI-IPent: An Active, Sterically Demanding Cross-Coupling Catalyst and Its Application in the Synthesis of Tetra-Ortho-Substituted Biaryls
Organ MG, et al.
Angewandte Chemie (International Edition in English), 48(13), 2383-2387 (2009)

Articles

Professor Mike Organ and co-workers have developed the PEPPSI™ (Pyridine-Enhanced Precatalyst Preparation Stabilization and Initiation) precatalysts for palladium-catalyzed cross-coupling reactions.

All of the preformed catalysts used in the kit are air and moisture stable complexes in their commercially available form.

Tools aid in kit setup for organic chemistry techniques, ensuring ease and success.

Tools aid in kit setup for organic chemistry techniques, ensuring ease and success.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service