Skip to Content
Merck
All Photos(1)

Key Documents

909114

Sigma-Aldrich

Methyl germanane

Synonym(s):

Methyl-germylidyne, Methyl-terminated germanane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
GeCH3
CAS Number:
Molecular Weight:
87.67
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23

form

solid

SMILES string

[Ge]C

InChI

1S/CH3Ge/c1-2/h1H3

InChI key

LCTMIXFKOJXTHA-UHFFFAOYSA-N

Application

Germanane is novel 2D graphene analog material. However, in sharp contrast to graphene it exhibits semiconducting properties with direct band gap of 1.50 eV. Germanane exhibits high electron mobility and its alkylation allows band gap tuning from 1.50 eV for allyl to 1.62 for methyl germanane. The semiconducting properties of alkylated germanane make them suitable for application is photocatalysis, optoelectronic, sensing etc

Storage and Stability

Air sensitive Material: Handle in glove box. Handle and store under nitrogen atmosphere

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liquid-Phase Exfoliation of Germanane Based on Hansen Solubility Parameters.
Nakamura D, et al.
Chemistry of Materials, 30 (15), 5333-5338 (2018)
Tailoring the Electronic Structure of Covalently Functionalized Germanane via the Interplay of Ligand Strain and Electronegativity.
Jiang S, et al.
Chemistry of Materials, 28, 8071-8071 (2016)
Electronic properties of germanane field-effect transistors.
Madhushankar B N, et al.
2d materials, 4, 021009-021009 (2017)
Zhenhua Liu et al.
Chemical communications (Cambridge, England), 50(75), 11046-11048 (2014-08-08)
A novel 2D material germanane (GeH), which was synthesized by an ion-exchange approach, was firstly found to exhibit photocatalytic performance in the hydrogen evolution of water splitting and decomposition of organic contaminants under illumination of visible light (λ ≥ 420
Andrew C Serino et al.
ACS nano, 11(8), 7995-8001 (2017-08-02)
The high theoretical energy density of alloyed lithium and germanium (Li15Ge4), 1384 mAh/g, makes germanium a promising anode material for lithium-ion batteries. However, common alloy anode architectures suffer from long-term instability upon repetitive charge-discharge cycles that arise from stress-induced degradation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service