Skip to Content
Merck
All Photos(1)

Key Documents

484385

Sigma-Aldrich

Gold(III) chloride solution

99.99% trace metals basis, 30 wt. % in dilute HCl

Synonym(s):

Auric chloride, Gold trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HAuCl4
CAS Number:
Molecular Weight:
339.79
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
liquid

Quality Level

Assay

99.99% trace metals basis

form

liquid

composition

Au, 17 wt. %

concentration

30 wt. % in dilute HCl

impurities

≤150.0 ppm Trace Metal Analysis

density

1.637 g/mL at 25 °C

SMILES string

[H][Au](Cl)(Cl)(Cl)Cl

InChI

1S/Au.4ClH/h;4*1H/q+3;;;;/p-3

InChI key

VDLSFRRYNGEBEJ-UHFFFAOYSA-K

General description

Gold(III) chloride solution, commonly called asauric chloride or auric acid is widely used as a precursor to synthesize goldcomplexes and stable gold nanoparticles. It also finds application in the fieldof catalysis.

Application

Gold(III) chloride solution can be used:
  • As a starting material to prepare thiolate-stabilized gold nanoparticles.
  • As an efficient catalyst for single-pot oxidative functionalization of alkanes and alcohols.
  • As a catalyst for diastereoselective alkylation reactions with chiral benzylic acetates.
Suitable precursormaterial for the synthesis of Gold nanorods by using Seed-Mediated Growth Methodwhich has been used for cancer cell imaging and photothermal therapy.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Golden Pathway to Thiolate-Stabilized Nanoparticles: Following the Formation of Gold(I) Thiolate from Gold(III) Chloride
Brian M. Barngrover and Christine M. Aikens
Journal of the American Chemical Society, 134, 12590-12595 (2022)
Preparation and Growth Mechanism of Gold Nanorods (NRs) Using Seed-Mediated Growth Method
Babak Nikoobakht, et al.
Chemistry of Materials, 15, 1957-1962 (2003)
Nanoparticle Targeting at Cells.
de la Fuente JM, et al.
Langmuir, 22, 3286-3293 (2006)
Commercial gold(I) and gold(III) compounds supported on carbon materials as greener catalysts for the oxidation of alkanes and alcohols
Sonia A. C. Carabineiro, et al.
ChemCatChem, 10, 1804-1813 (2018)
Gold(III)Chloride-Catalyzed Diastereoselective Alkylation Reactions with Chiral Benzylic Acetates
Philipp Rubenbauer and Thorsten Bach
advanced synthesis and catalysis, 350, 1125-1130 (2008)

Articles

Single molecule electronics is the endeavour of constructing electronic circuitry with single molecules as the fundamental building block.

Single molecule electronics is the endeavour of constructing electronic circuitry with single molecules as the fundamental building block.

Single molecule electronics is the endeavour of constructing electronic circuitry with single molecules as the fundamental building block.

Single molecule electronics is the endeavour of constructing electronic circuitry with single molecules as the fundamental building block.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service