Skip to Content
Merck
All Photos(1)

Documents

907081

Sigma-Aldrich

Methacrylamide poly(ethylene glycol) amine hydrochloride

average Mn 400, cross-linking reagent polymerization reactions

Synonym(s):

Methacrylamide poly(ethylene glycol) amine, PEG amine monomethacrylamide, PEG diamine mono methacrylamide, PEG methacrylamide, PEG monomethacrylamide, Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H10NO(C2H4O)nNH2 · HCl
UNSPSC Code:
12352101
NACRES:
NA.23

product name

Methacrylamide poly(ethylene glycol) amine hydrochloride, average Mn 400

form

liquid

mol wt

average Mn 400 (by NMR)
average Mn 400

reaction suitability

reagent type: cross-linking reagent
reaction type: Polymerization Reactions

color

pale yellow

polymer architecture

shape: linear
functionality: heterobifunctional

storage temp.

2-8°C

Application

PEG for cross-linking. PEGylated materials have found broad use in drug delivery systems, virology, and immunology, as the incorporation of PEG improves pharmacological properties such as increased water solubility, enhanced resistance to degradation (protein hydrolysis), increased circulation half-life, and reduced antigenicity. Polymer for preparing enzyme conjugates soluble in organic solvents; promising drug carrier.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun-ichi Furukawa et al.
Analytical chemistry, 80(4), 1094-1101 (2008-01-22)
Changes in protein glycosylation profoundly affect protein function. To understand these effects of altered protein glycosylation, we urgently need high-throughput technologies to analyze glycan expression and glycan-protein interactions. Methods are not available for amplification of glycans; therefore, highly efficient sample
Hideyuki Shimaoka et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(6), 1664-1673 (2007-01-17)
The development of rapid and efficient methods for high-throughput protein glycomics is of growing importance because the glycoform-focused reverse proteomics/genomics strategy will greatly contribute to the discovery of novel biomarkers closely related to cellular development, differentiation, growth, and aging as
Emerson Giovanelli et al.
Langmuir : the ACS journal of surfaces and colloids, 28(43), 15177-15184 (2012-09-26)
High colloidal stability in aqueous conditions is a prerequisite for fluorescent nanocrystals, otherwise known as "quantum dots", intended to be used in any long-term bioimaging experiment. This essential property implies a strong affinity between the nanoparticles themselves and the ligands

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service