Skip to Content
Merck
All Photos(2)

Key Documents

903000

Sigma-Aldrich

[4,4′-Bis(1,1-dimethylethyl)-2,2′-bipyridine] nickel (II) dichloride

Synonym(s):

(4,4′-dtbbpy)NiCl2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24Cl2N2Ni
CAS Number:
Molecular Weight:
398.00
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

form

powder or crystals

reaction suitability

core: nickel
reaction type: Cross Couplings
reagent type: catalyst

mp

>300 °C

SMILES string

CC(C1=CC(C2=CC(C(C)(C)C)=CC=N2)=NC=C1)(C)C.Cl[Ni]Cl

InChI key

PCWIKFRTCXESOT-UHFFFAOYSA-L

Application

[4,4′-Bis(1,1-dimethylethyl)-2,2′-bipyridine] nickel (II) dichloride can be used as a catalyst in:
  • Decarboxylative arylation of oxo acids.
  • Acylation of ethers.
  • Cross-coupling of aryl bromides with alcohols.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photocatalytic α-Acylation of Ethers
Sun Z, et al.
Organic Letters, 19, 3727-3730 (2017)
Aryl Ketones as Single-Electron-Transfer Photoredox Catalysts in Nickel-Catalyzed the Homocoupling of Aryl Halides
Masuda Y, et al.
European Journal of Organic Chemistry, 5822-5825 (2016)
Martins S Oderinde et al.
The Journal of organic chemistry, 80(15), 7642-7651 (2015-07-04)
In order to achieve reproducibility during iridium-photoredox and nickel dual-catalyzed sp(3)-sp(2) carbon-carbon bond-forming reactions, we investigated the role that molecular oxygen (O2), solvent and light-source (CF lamp or blue LED) play in a variety of Ir-photoredox mediated transformations. The presence
Merging photoredox and nickel catalysis: The direct synthesis of ketones by the decarboxylative arylation of α-oxo acids
Chu L, et al.
Angewandte Chemie (International ed. in English), 127, 8040-8044 (2015)
Ir III/Ni II-Metallaphotoredox catalysis: the oxidation state modulation mechanism versus the radical mechanism
Zhu B, et al.
Chemical Communications (Cambridge, England), 54, 5968-5971 (2018)

Articles

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service