Skip to Content
Merck
All Photos(2)

Key Documents

901409

Sigma-Aldrich

[Ir(dF(Me)ppy)2(dtbbpy)]PF6

Synonym(s):

Iridium(III) bis[2-(2,4-difluorophenyl)-5-methylpyridine-N,C20]-4,40-di-tert-butyl-2,20-bipyridine hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H40F10IrN4P
CAS Number:
Molecular Weight:
1013.97
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

core: iridium
reaction type: Photocatalysis
reagent type: catalyst

photocatalyst activation

450 nm

InChI key

XAJPUJWOMJJPNU-UHFFFAOYSA-N

Application

[Ir(dF(Me)ppy)2(dtbbpy)]PF6 is a photocatalyst readily facilitates hydroamination of olefins as well as the decarboxylative arylation and vinylation of carboxylic acids via metallaphotoredox catalysis.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Correlating electronic structures to electrochemiluminescence of cationic Ir complexes.
Swanick KN, et al.
Royal Society of Chemistry Advances, 43, 19961-19964 (2013)
Victoria Dimakos et al.
Journal of the American Chemical Society, 141(13), 5149-5153 (2019-03-23)
Diphenylborinic acid serves as a cocatalyst for site- and stereoselective C-H alkylation reactions of carbohydrates under photoredox conditions using quinuclidine as the hydrogen atom transfer mediator. Products arising from selective abstraction of the equatorial hydrogens of cis-1,2-diol moieties, followed by
Sébastien Ladouceur et al.
Inorganic chemistry, 50(22), 11514-11526 (2011-10-04)
Herein we report the synthesis of 4-aryl-1-benzyl-1H-1,2,3-triazoles (atl), made via "Click chemistry" and their incorporation as cyclometallating ligands into new heteroleptic iridium(III) complexes containing diimine (N(^)N) ancillary ligands 2,2'-bipyridine (bpy) and 4,4'-di-tert-butyl-2,2'-bipyridine (dtBubpy). Depending on decoration, these complexes emit from
Andrew J Musacchio et al.
Science (New York, N.Y.), 355(6326), 727-730 (2017-02-18)
The intermolecular hydroamination of unactivated alkenes with simple dialkyl amines remains an unsolved problem in organic synthesis. We report a catalytic protocol for efficient additions of cyclic and acyclic secondary alkyl amines to a wide range of alkyl olefins with
Strongly Blue Luminescent Cationic Iridium(III) Complexes with an Electron-Rich Ancillary Ligand: Evaluation of Their Optoelectronic and Electrochemiluminescence Properties.
Ladouceur S, et al.
European Journal of Inorganic Chemistry, 2013(30), 5329-5343 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service