Skip to Content
Merck
All Photos(1)

Key Documents

901802

Sigma-Aldrich

Fluorescent nanodiamond

nitrogen vacancy ~3 ppm, 140 nm avg. part. size, amine functionalized, PEG 3000 coated, 1 mg/mL in deionized water

Synonym(s):

Fluorescent nanoparticles, Nanodiamonds

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
NACRES:
NA.23

form

nanoparticles

composition

nitrogen vacancy, ~3 ppm

concentration

1 mg/mL in deionized water

avg. part. size

140 nm

Looking for similar products? Visit Product Comparison Guide

Application

Nanodiamonds (NDs) containing NV color centers exhibit fluorescence in red/near infra-red (NIR) region. The fluorescence in NDs originates from nitrogen vacancies and, unlike quantum dots and organic dyes, NDs do not photobleach. Fluorescent NDs find applications in background-free imaging, flow cytometry, super-resolution imaging etc. Our PEG coated aminated fluorescent nanodiamonds are well suited for conjugation with activated biomolecules where steric hindrance is of concern.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly Fluorescent Nanodiamonds Protein-Functionalized for Cell Labeling and Targeting.
B. Chang et al.,
Advances in Functional Materials, 23, 5737-5737 (2013)
Functionality is Key: Recent Progress in the Surface Modification of Nanodiamond.
A. Krueger and D. Lang
Advances in Functional Materials, 22, 890-890 (2012)
Nanodiamond bioconjugate probes and their collection by electrophoresis.
S. C. Hens et al.,
Diamond and Related Materials, 17, 1858-1858 (2008)
Nanodiamond-mediated drug delivery and imaging: Challenges and opportunities.
V. Vaijayanthimala et al.,
Expert Opinion on Drug Delivery, 12, 1-1 (2014)
Yuen Yung Hui et al.
Optics express, 18(6), 5896-5905 (2010-04-15)
Dynamics of fluorescent diamond nanoparticles in HeLa cells has been studied with two-photon fluorescence correlation spectroscopy (FCS). Fluorescent nanodiamond (FND) is an excellent fluorescent probe for bioimaging application, but they are often trapped in endosomes after cellular uptake. The entrapment

Articles

Fluorescent Nanodiamond Particles (FNDs)- Find properties and applications of nanodiamond particles.

Fluorescent Nanodiamonds and biofunctionalization strategies are detailed for imaging and bioconjugation applications.

Fluorescent Nanodiamonds and biofunctionalization strategies are detailed for imaging and bioconjugation applications.

Fluorescent Nanodiamonds and biofunctionalization strategies are detailed for imaging and bioconjugation applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service