Skip to Content
Merck
All Photos(1)

Key Documents

900755

Sigma-Aldrich

SLAP TA

95%

Synonym(s):

3-(((Trimethylsilyl)methyl)thio)propan-1-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H19NSSi
CAS Number:
Molecular Weight:
177.38
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

density

0.90452

functional group

amine
thioether

storage temp.

2-8°C

SMILES string

[Si](CSCCCN)(C)(C)C

InChI

1S/C7H19NSSi/c1-10(2,3)7-9-6-4-5-8/h4-8H2,1-3H3

InChI key

CNESDLLESCAVSR-UHFFFAOYSA-N

Application

When used with aliphatic or aromatic aldehydes, this silicon amine protocol SLAP reagent enables the photomediated synthesis of thiomorpholines and thiazepanes. For photocatalytic cross-coupling with this SLAP Reagent, Ir[(ppy)2dtbbpy]PF6 (747769) is used with a recommended starting combination of Lewis acids Bi(OTf)3 (633305) and Cu(OTf)2 (283673) prior to subsequent substrate-specific optimization. This product was introduced in collaboration with the Bode Research Group and provides a tin-free alternative to SnAP (tin amine protocol) reagents and is well-suited for scale-up reactions.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.0 °F

Flash Point(C)

23.89 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Moritz K Jackl et al.
Organic letters, 19(17), 4696-4699 (2017-08-17)
Photocatalytic coupling of aldehydes and silicon amine protocol (SLAP) reagents enables the simple, scalable synthesis of substituted morpholines, oxazepanes, thiomorpholines, and thiazepanes under continuous flow conditions. Key to the success of this process is the combination of an inexpensive organic
Sheng-Ying Hsieh et al.
Organic letters, 18(9), 2098-2101 (2016-04-22)
Silicon amine protocol (SLAP) reagents for photocatalytic cross-coupling with aldehydes and ketones to form N-unprotected piperazines have been developed. This blue light promoted process tolerates a wide range of heteroaromatic, aromatic, and aliphatic aldehydes and structurally and stereochemically complex SLAP
Sheng-Ying Hsieh et al.
ACS central science, 3(1), 66-72 (2017-02-06)
Redox neutral photocatalytic transformations often require careful pairing of the substrates and photoredox catalysts in order to achieve a catalytic cycle. This can limit the range of viable transformations, as we recently observed in attempting to extend the scope of

Protocols

The expanding class of SLAP reagents provides access to saturated N-heterocycles and aliphatic aldehydes and ketones without additional reagents or the generation of toxic byproducts.

The expanding class of SLAP reagents provides access to saturated N-heterocycles and aliphatic aldehydes and ketones without additional reagents or the generation of toxic byproducts.

The expanding class of SLAP reagents provides access to saturated N-heterocycles and aliphatic aldehydes and ketones without additional reagents or the generation of toxic byproducts.

The expanding class of SLAP reagents provides access to saturated N-heterocycles and aliphatic aldehydes and ketones without additional reagents or the generation of toxic byproducts.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service