Skip to Content
Merck
All Photos(2)

Key Documents

88005

Sigma-Aldrich

Tetrapentylammonium hydroxide solution

~20% in H2O (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)4]4N(OH)
CAS Number:
Molecular Weight:
315.58
Beilstein:
6785584
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

form

solution

concentration

~20% in H2O (T)

impurities

≤0.05% halides (as bromide)

anion traces

sulfate (SO42-): ≤500 mg/kg

functional group

amine

SMILES string

[OH-].CCCCC[N+](CCCCC)(CCCCC)CCCCC

InChI

1S/C20H44N.H2O/c1-5-9-13-17-21(18-14-10-6-2,19-15-11-7-3)20-16-12-8-4;/h5-20H2,1-4H3;1H2/q+1;/p-1

InChI key

JVOPCCBEQRRLOJ-UHFFFAOYSA-M

Application

Tetrapentylammonium hydroxide can be used:
  • As a model compound in the studies of the effects of quaternary ammonium salts on the formation of clathrate hydrates of methane.
  • To maintain the pH of the aqueous phase to study the adsorption dynamics of nanoparticles at fluid interfaces.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Semiclathrate hydrates of methane+ tetraalkylammonium hydroxides
Su Y, et al.
Fuel: The Science and Technology of Fuel and Energy, 203, 618-626 (2017)
Nanoparticle adsorption dynamics at fluid interfaces
Hua X, et al.
Soft Matter, 14(19), 3818-3828 (2018)
Xiaoqing Hua et al.
Langmuir : the ACS journal of surfaces and colloids, 34(16), 4830-4842 (2018-04-11)
Nanoparticles (NPs) can add functionality (e.g., catalytic, optical, rheological) to an oil-water interface. Adsorption of ∼10 nm NPs can be reversible; however, the mechanisms for adsorption and its effects on surface pressure remain poorly understood. Here we demonstrate how the
Maria G Signorello et al.
European journal of biochemistry, 270(9), 2005-2012 (2003-04-24)
The results of the present study show that human platelets take up l-arginine by two transport systems which are compatible with the systems y+ and y+L. These Na+independent transporters have been distinguished by treating platelets with N-ethylmaleimide that blocks selectively
Stefan Verlohren et al.
Hypertension (Dallas, Tex. : 1979), 44(3), 271-276 (2004-08-11)
Periadventitial adipose tissue produces vasoactive substances that influence vascular contraction. Earlier studies addressed this issue in aorta, a vessel that does not contribute to peripheral vascular resistance. We tested the hypothesis that periadventitial adipose tissue modulates contraction of smaller arteries

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service