Skip to Content
Merck
All Photos(1)

Key Documents

86909

Sigma-Aldrich

Tetrabutylammonium nonafluorobutanesulfonate

≥98.0%

Synonym(s):

Perfluorobutanesulfonic acid tetrabutylammonium salt, Tetrabutylammonium perfluorobutanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(CF3CF2CF2CF2SO3)
CAS Number:
Molecular Weight:
541.56
Beilstein:
5219877
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

form

powder

mp

50-53 °C

SMILES string

CCCC[N+](CCCC)(CCCC)CCCC.[O-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C16H36N.C4HF9O3S/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;5-1(6,3(9,10)11)2(7,8)4(12,13)17(14,15)16/h5-16H2,1-4H3;(H,14,15,16)/q+1;/p-1

InChI key

VECGWISURDHBJL-UHFFFAOYSA-M

General description

Tetrabutylammonium nonafluorobutanesulfonate is a fluorinated ionic liquid (FIL). Its solid-fluid transition and nanoscale structuring have been studied using differential scanning calorimetry (DSC), rheology techniques and molecular dynamic simulations.

Other Notes

Ionic liquid

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Which structural features stand behind micelization of ionic liquids? Quantitative Structure-Property Relationship studies
Barycki, Maciej, Anita Sosnowska, and Tomasz Puzyn
Journal of Colloid and Interface Science, 487, 475-483 (2017)
Validation of a screening method based on liquid chromatography coupled to high-resolution mass spectrometry for analysis of perfluoroalkylated substances in biota
Berger, Urs, and Marianne Hauk?s.
Journal of Chromatography A, 1081.2, 210-217 (2005)
(Liquid+ liquid) equilibria of perfluorocarbons with fluorinated ionic liquids
Martinho, S., et al.
The Journal of Chemical Thermodynamics, 64, 71-79 (2013)
M. Badri et al.
Tetrahedron Letters, 33, 4435-4435 (1992)
C.F. Poole et al. et al.
Molten Salt Techniques, 4, 41-41 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service