Skip to Content
Merck
All Photos(3)

Key Documents

562629

Sigma-Aldrich

Nonafluorobutane-1-sulfonic acid

97%

Synonym(s):

1,1,2,2,3,3,4,4,4-Nonafluoro-1-butanesulfonic acid, 1-Perfluorobutanesulfonic acid, Nonafluoro-1-butanesulfonic acid, Nonafluorobutan-1-sulfonic acid, Nonafluorobutanesulfonic acid, Perfluorobutanesulfonic acid, n-Nonafluorobutanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)3SO3H
CAS Number:
Molecular Weight:
300.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.3230 (lit.)

bp

112-114 °C/14 mmHg (lit.)

density

1.811 g/mL at 25 °C (lit.)

functional group

fluoro
sulfonic acid

storage temp.

2-8°C

SMILES string

OS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C4HF9O3S/c5-1(6,3(9,10)11)2(7,8)4(12,13)17(14,15)16/h(H,14,15,16)

InChI key

JGTNAGYHADQMCM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nonafluorobutane-1-sulfonic acid is a colorless liquid that can be considered as a superacid, which can be synthesized by reacting 1-iodononafluorobutane with sodium dithionite in the presence of sodium bicarbonate in acetonitrile/water.

Application

Nonafluorobutane-1-sulfonic acid may be used as catalyst in the synthesis of 6-chloro-6H-dibenz[c,e][1,2]oxaphosphorin and N-benzylpyridin-2-amine.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

350.6 °F - Pensky-Martens closed cup

Flash Point(C)

177 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Black D
Science of Synthesis: Houben-Weyl Methods of Molecular Transformations, 15 (2014)
"A general method for the preparation of perfluoroalkanesulfonyl chlorides"
Scott.H.J P, et al.
Journal of Fluorine Chemistry, 126(8), 1196-1201 (2005)
Lizhu Tang et al.
Chemosphere, 256, 127169-127169 (2020-05-29)
Parental exposure to perfluorobutane sulfonate (PFBS), an aquatic pollutant of emerging concern, is previously found to impair the embryonic development of offspring. However, the impairing mechanisms remain to clarify. In the present study, adult zebrafish were exposed to 0, 10
Karilyn E Sant et al.
Toxicological sciences : an official journal of the Society of Toxicology, 167(1), 258-268 (2018-09-22)
Following the phase-out of highly persistent perfluorosulfonates in the United States from non-stick and stain-resistant products in the early 2000s, perfluorobutanesulfonic acid (PFBS) has replaced these compounds as a primary surfactant. Measurements of PFBS in environmental and human samples have
Weipeng Qi et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 120, 340-345 (2018-07-22)
Perfluorobutanesulfonic acid (PFBS) is used as the replacement of perfluorooctanesulfonic acid (PFOS) since 2000 because of the concern on PFOS' persistence in the environment and the bioaccumulation in animals. Accumulating evidence has shown the correlation between the exposure to perfluorinated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service