Skip to Content
Merck
All Photos(3)

Key Documents

861324

Sigma-Aldrich

p-Rosolic acid

Dye content 84 %

Synonym(s):

4-[Bis(4-hydroxyphenyl)methylene]-2,5-cyclohexadienone, Aurin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14O3
CAS Number:
Molecular Weight:
290.31
Colour Index Number:
43800
Beilstein:
2055205
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

composition

Dye content, 84%

technique(s)

titration: suitable

pH

5.0-6.8, yellow to pink

cation traces

C: 66.0-90.8%

λmax

482 nm

ε (extinction coefficient)

≥11000 at 264-270 nm in ethanol
≥50000 at 478-486 nm in ethanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

O=C(C=C1)C=CC1=C(C2=CC=C(O)C=C2)C3=CC=C(O)C=C3

InChI

1S/C19H14O3/c20-16-7-1-13(2-8-16)19(14-3-9-17(21)10-4-14)15-5-11-18(22)12-6-15/h1-12,20-21H

InChI key

FYEHYMARPSSOBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

p-Rosolic acid (C.I. 43800) is an acid-baseindicator from the triphenylmethane group. It is also known as aurin and 4-[Bis(4-hydroxyphenyl)methylene]-2,5-cyclohexadienone.

Application

p-Rosolic acid has been used to induce cellular stress in HEK293 cells.

Biochem/physiol Actions

The transition range of p-Rosolic acid is around pH 6.6-8.0 (Below this pH range it is yellow and above it red).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Simultaneous detection of the subcellular localization of RNAs and proteins in cultured cells by combined multicolor RNA-FISH and IF.
Meyer C, et al.
Methods, S1046-2023 (2016)
A Batistatou et al.
The Journal of cell biology, 122(3), 523-532 (1993-08-01)
Serum-free PC12 cell cultures have been used to study the mechanisms of neuronal death after neurotrophic factor deprivation. We previously reported that PC12 cells undergo "apoptotic" internucleosomal DNA cleavage after withdrawal of trophic support. Here, we have used a sensitive
[Not Available].
F PASCUCCI et al.
Rivista critica di clinica medica, 45(6-9), 484-493 (1945-11-30)
[Not Available].
A DESAUX et al.
Annales de dermatologie et de syphiligraphie, 6(9), 504-504 (1946-09-01)
J Chen et al.
Thrombosis research, 66(2-3), 111-120 (1992-05-01)
To determine whether heparin potentiation of platelet aggregation is related to platelet GP IIb/IIIa and GP Ib receptors, four series of experiments were performed on blood from normal volunteers. In the first experiment pretreatment with the monoclonal antibody 7E3 (MAb

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service