79874
2-Chloro-1,3,5-trinitrobenzene
≥98.0% (HPLC)
Synonym(s):
2,4,6-Trinitrochlorobenzene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
≥98.0% (HPLC)
form
lumps
functional group
chloro
nitro
SMILES string
[O-][N+](=O)c1cc(c(Cl)c(c1)[N+]([O-])=O)[N+]([O-])=O
InChI
1S/C6H2ClN3O6/c7-6-4(9(13)14)1-3(8(11)12)2-5(6)10(15)16/h1-2H
InChI key
HJRJRUMKQCMYDL-UHFFFAOYSA-N
Other Notes
moistened with ~15% water
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Sol. 1
Supplementary Hazards
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Toxicology letters, 213(3), 392-401 (2012-07-31)
Immunosuppressive environmental chemicals may exacerbate allergic diseases, including atopic dermatitis (AD). We examined the effects of the immunosuppressive environmental chemicals methoxychlor, parathion, piperonyl butoxide, dexamethasone, and cyclophosphamide on picryl-chloride-induced AD in NC/Nga mice. Mice were orally exposed (age, 5 weeks)
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons, 17(1), 60-68 (2016-08-09)
Understanding the evolution of the direct and indirect pathways of allorecognition following tissue transplantation is essential in the design of tolerance-promoting protocols. On the basis that donor bone marrow-derived antigen-presenting cells are eliminated within days of transplantation, it has been
Journal of ethnopharmacology, 145(1), 294-302 (2012-11-15)
Korean red ginseng (KRG) has been shown to possess various biological activities including anti-inflammatory properties. We aimed to investigate the effects and mechanism of KRG on the prevention of atopic dermatitis (AD) using a mouse model. The effect of KRG
Chemical research in toxicology, 24(11), 2018-2027 (2011-10-26)
The skin sensitization potency of chemicals is partly related to their reactivity to proteins. This can be quantified as the rate constant of the reaction with a model peptide, and a kinetic profiling approach to determine rate constants was previously
International immunopharmacology, 11(10), 1628-1632 (2011-06-07)
A chymase inhibitor SUN13834 has been shown to improve skin condition in animal models for atopic dermatitis. In the present study, effective dosages of SUN13834 for atopic dermatitis patients were predicted by pharmacokinetic/pharmacodynamic (PK/PD) analyses of SUN13834 in NC/Nga mice
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service