769770
Resomer® RG 750 S, Poly(D,L-lactide-co-glycolide)
ester terminated, lactide:glycolide 75:25
Synonym(s):
Resomer® RG 750 S, PLGA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HO[C2H4O2]n[C2H2O2]mCH3
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Recommended Products
Quality Level
form
amorphous
feed ratio
lactide:glycolide 75:25
viscosity
0.8-1.2 dL/g
storage temp.
2-8°C
SMILES string
O2C(C(=O)OC(C2=O)C)C.O1CC(=O)OCC1=O
InChI
1S/C6H8O4.C4H4O4/c1-3-5(7)10-4(2)6(8)9-3;5-3-1-7-4(6)2-8-3/h3-4H,1-2H3;1-2H2
InChI key
LCSKNASZPVZHEG-UHFFFAOYSA-N
Application
Resomer® RG 750 S, Poly(D,L-lactide-co-glycolide), a biodegradable and biocompatible aliphatic polyester, can be used in the development of nano and microparticles. It is a pharmaceutical polymer that can potentially be utilized in drug delivery and gene delivery. It forms a three-dimensional porous scaffold, which can be used in tissue engineering.
Legal Information
RESOMER is a registered trademark of Evonik Rohm GmbH
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Development and evaluation of novel biodegradable microspheres based on poly (d, l-lactide-co-glycolide) and poly (varepsilon-caprolactone) for controlled delivery of doxycycline in the treatment of human periodontal pocket: in vitro and in vivo studies
Mundargi RC, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 119(1), 59-68 (2007)
In vitro degradation of three-dimensional porous poly (D, L-lactide-co-glycolide) scaffolds for tissue engineering
Wu L and Ding J
Biomaterials, 25(27), 5821-5830 (2004)
Nano/micro technologies for delivering macromolecular therapeutics using poly (D, L-lactide-co-glycolide) and its derivatives
Mundargi RC, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 125(3), 193-209 (2008)
Nanoparticles made of fluorescence-labelled poly (L-lactide-co-glycolide): preparation, stability, and biocompatibility
Weiss B, et al.
Journal of Nanoscience and Nanotechnology, 6(9-10), 3048-3056 (2006)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service