Skip to Content
Merck
All Photos(1)

Documents

739944

Sigma-Aldrich

Resomer® RG 504, Poly(D,L-lactide-co-glycolide)

lactide:glycolide 50:50, ester terminated, Mw 38,000-54,000

Synonym(s):

PLGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C3H4O2]x[C2H2O2]y
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

amorphous

feed ratio

lactide:glycolide 50:50

mol wt

Mw 38,000-54,000

degradation timeframe

<3 months

viscosity

0.45-0.60 dL/g, 0.1 % (w/v) in chloroform(25 °C, Ubbelohde) (size 0c glass capillary viscometer)

transition temp

Tg 46-50 °C

storage temp.

2-8°C

InChI

1S/C6H8O4.C4H4O4/c1-3-5(7)10-4(2)6(8)9-3;5-3-1-7-4(6)2-8-3/h3-4H,1-2H3;1-2H2

InChI key

LCSKNASZPVZHEG-UHFFFAOYSA-N

Application

Resomer® RG 504, Poly(ᴅ,ʟ-lactide-co-glycolide) can be used to prepare polymeric nanoparticles that are effective carriers for anti-cancer natural substances.

Poly(D, L-lactide-co-glycolide)microspheres can be used to fabricate a hemostatic delivery device forthrombin.

Features and Benefits

Controlled release of bioactive agents, sutures and bioabsorbable implantable devices.

Legal Information

Product of Evonik
RESOMER is a registered trademark of Evonik Rohm GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ananthakrishnan Soundaram Jeevarathinam et al.
Angewandte Chemie (International ed. in English) (2019-12-17)
We report a new approach to monitor drug release from nanocarriers via a paclitaxel-methylene blue conjugate (PTX-MB) with redox activity. This construct is in a photoacoustically silent reduced state inside poly(lactic-co-glycolic acid) (PLGA) nanoparticles (PTX-MB@PLGA NPs). During release, PTX-MB is
Rongcai Liang et al.
International journal of pharmaceutics, 454(1), 344-353 (2013-07-23)
Peptide or protein degradation often occurs when water flows into the dosage form. The aim of this study was to investigate the effect of water on exenatide acylation in poly(lactide-co-glycolide) (PLGA) microspheres. Exenatide-loaded PLGA microspheres were incubated at different relative
Igor Jeroukhimov et al.
Journal of the American College of Surgeons, 218(1), 102-107 (2013-11-12)
Chronic pain after inguinal hernia repair occurs in 16% to 62% of patients. The underlying mechanism probably involves sensory nerve damage and abnormal healing that might be influenced by the materials chosen for the procedure. We hypothesize that nonabsorbable sutures
Maria Kirzhner et al.
Ophthalmology, 120(6), 1300-1304 (2013-02-13)
To compare wrapped and polymer-coated hydroxyapatite implants in children undergoing primary enucleation with no adjuvant therapies. Retrospective, interventional cohort study. All children undergoing primary enucleation without adjuvant therapies between 1999 and 2009 at a tertiary pediatric cancer hospital. Review and
Teresa Musumeci et al.
International journal of pharmaceutics, 440(2), 135-140 (2012-10-20)
Melatonin, a neurohormone secreted by the pineal gland, is able to modulate intraocular pressure (IOP). The aim of this study was to generate nanoparticle (NPs) sustained release formulations that allow to extend the pre-corneal residence time of melatonin, thus prolonging

Articles

Interest in utilizing biodegradable polymers for biomedical applications has grown since the 1960s.

Synthetic aliphatic polyesters dominate resorbable biomaterials in clinical use.

Synthetic aliphatic polyesters dominate resorbable biomaterials in clinical use.

Synthetic aliphatic polyesters dominate resorbable biomaterials in clinical use.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service