Merck
  • Evaluation of detection sensitivity in comprehensive two-dimensional liquid chromatography separations of an active pharmaceutical ingredient and its degradants.
Evaluation of detection sensitivity in comprehensive two-dimensional liquid chromatography separations of an active pharmaceutical ingredient and its degradants.
Analytical and bioanalytical chemistry (2014-07-30)
Dwight R Stoll, Eric S Talus, David C Harmes, Kelly Zhang
ABSTRACT
In this paper, we describe the findings of a study aimed at assessing the detection sensitivity of comprehensive two-dimensional high-performance liquid chromatography (LCxLC) separation of a degraded active pharmaceutical ingredient (API) with UV absorption as the detection technique. Specifically, we have examined the impact of the volume and solvent composition (referred to as "interface conditions") of fractions of first-dimension column effluent transferred to the second dimension for further separation on the ability to resolve and detect low-abundance compounds. Historically, LCxLC has been perceived as being inferior to 1D-LC from the point of view of detection sensitivity. In this work, we demonstrate that LCxLC is sufficiently sensitive to be useful in the pharmaceutical context where in general impurities present at 0.05 % (relative to the API concentration) should be quantified. Moreover, we find that this level of sensitivity is only attained under certain conditions: dilution of the first column effluent with weak solvent (water in this case) prior to injection into the second-dimension column is very beneficial because it promotes focusing of the analyte band in the second column, thereby improving the detection sensitivity of the LCxLC system; and, quantitation limits are also a strong function of peak location in the second-dimension separation window, where baseline disturbances near the dead time of the second column can limit reliable detection of low-abundance compounds.
Sigma-Aldrich
Butylbenzene, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Butylbenzene, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Bromobenzene, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Bromobenzene, ReagentPlus®, 99%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ultrapure Acetonitrile
SKU
Pack Size
Availability
Price
Quantity
Supelco
Acetonitrile, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Naproxen, VETRANAL®, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Naproxen, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Supelco
Perchloric acid, 0.01 M HClO4 in water (0.01N), eluent for IC
SKU
Pack Size
Availability
Price
Quantity
Supelco
Naproxen, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
SKU
Pack Size
Availability
Price
Quantity
USP
Naproxen, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Perchloric acid, ACS reagent, 70%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Perchloric acid, ACS reagent, 60%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Perchloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 70.0-72.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Perchloric acid, puriss. p.a., ACS reagent, packed in coated, shock- and leak-protected glass bottle, ≥60% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)