Skip to Content
Merck
All Photos(1)

Documents

733326

Sigma-Aldrich

cis-4,7,10,13,16,19-Docosahexaenoic acid-21,21,22,22,22-d5

≥98 atom % D, ≥98% (CP)

Synonym(s):

DHA-d5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22D5H27O2
CAS Number:
Molecular Weight:
333.52
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.24

isotopic purity

≥98 atom % D

Assay

≥98% (CP)

form

liquid

mass shift

M+5

shipped in

dry ice

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])C([2H])([2H])\C=C/C\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCC(O)=O

InChI

1S/C22H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h3-4,6-7,9-10,12-13,15-16,18-19H,2,5,8,11,14,17,20-21H2,1H3,(H,23,24)/b4-3-,7-6-,10-9-,13-12-,16-15-,19-18-/i1D3,2D2

InChI key

MBMBGCFOFBJSGT-RPBOKJFVSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer T Wolstenholme et al.
Alcoholism, clinical and experimental research (2018-05-23)
Complex interactions between environmental and genetic factors influence the risk of developing alcohol use disorder (AUD) in humans. To date, studies of the impact of environment on AUD risk have primarily focused on psychological characteristics or on the effects of
Valentina Spigoni et al.
Food & function, 5(8), 1881-1890 (2014-06-20)
Recent data suggest that n-3 PUFA exert beneficial effects on endothelial progenitor cell (EPC) biology. We sought to investigate whether these effects might be mediated by enhanced EPC in vitro function and/or in vivo bioavailability. CACs and late-outgrowth EPCs were
Melissa McDougall et al.
Free radical biology & medicine, 110, 250-260 (2017-06-25)
Vitamin E (α-tocopherol; VitE) is a lipophilic antioxidant required for normal embryonic development in vertebrates, but the long-term effects of embryonic VitE deficiency, and whether they are ameliorated by feeding VitE-adequate diets, remain unknown. We addressed these questions using a
Junjie Zhang et al.
Talanta, 222, 121679-121679 (2020-11-11)
The identification of new biomarkers (e.g., metabolic biomarkers) will facilitate not only the diagnosis of stroke but also the differentiation of stroke subtypes, especially the discrimination of ischaemic stroke from intracerebral hemorrhage. Herein, we develop for the first time an
Subhadip Senapati et al.
Biochimica et biophysica acta, 1860(6), 1403-1413 (2018-04-08)
Docosahexaenoic acid (DHA) is enriched in photoreceptor cell membranes. DHA deficiency impairs vision due to photoreceptor cell dysfunction, which is caused, at least in part, by reduced activity of rhodopsin, the light receptor that initiates phototransduction. It is unclear how

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service