Skip to Content
Merck
All Photos(2)

Key Documents

709298

Sigma-Aldrich

CX32

Umicore, 95%

Synonym(s):

Chlorophenylallyl[1,3-bis(2,6-diisopropylphenyl)-2-imidazolidinylidene]palladium(II), [1,3-Bis(2,6-di-isopropylphenyl)-4,5-dihydroimidazol-2-ylidene]chloro][3-phenylallyl]palladium(II), [1,3-Bis[2,6-bis(1-methylethyl)phenyl]-2-imidazolidinylidene]chloro[(1,2,3-η)-3-phenyl-2-propenyl]palladium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H47ClN2Pd
CAS Number:
Molecular Weight:
649.64
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

127-130 °C

SMILES string

[CH2][CH][CH]c1ccccc1.CC(C)c2cccc(C(C)C)c2N3CCN(\C3=[Pd]/Cl)c4c(cccc4C(C)C)C(C)C

InChI

1S/C27H38N2.C9H9.ClH.Pd/c1-18(2)22-11-9-12-23(19(3)4)26(22)28-15-16-29(17-28)27-24(20(5)6)13-10-14-25(27)21(7)8;1-2-6-9-7-4-3-5-8-9;;/h9-14,18-21H,15-16H2,1-8H3;2-8H,1H2;1H;/q;;;+1/p-1

InChI key

KEHHNXMVROYFNY-UHFFFAOYSA-M

Application

Catalyst for:
  • Heterocyclization reactions
  • Buchwald-Hartwig cross coupling
  • Suzuki-Miyaura reactions

Legal Information

Product of Umicore

Additional information available at www.pmc.umicore.com

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service