Skip to Content
Merck
All Photos(1)

Key Documents

72243

Sigma-Aldrich

Nickel(II) bromide trihydrate

98% (AT)

Synonym(s):

Nickel dibromide hydrate, Nickelous bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiBr2 · 3H2O
CAS Number:
Molecular Weight:
272.55
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
crystals

grade

for analytical purposes

Quality Level

Assay

98% (AT)

form

crystals

reaction suitability

reagent type: catalyst
core: nickel

application(s)

battery manufacturing

SMILES string

O.O.O.Br[Ni]Br

InChI

1S/2BrH.Ni.3H2O/h2*1H;;3*1H2/q;;+2;;;/p-2

InChI key

UQPSGBZICXWIAG-UHFFFAOYSA-L

Application

  • Development of di and polynuclear lead (II) salen (or reduced salen) complexes having PbO2M (M= Cu/Ni) cores: Synthetic strategies and structures: Investigates the synthesis and structural features of polynuclear lead(II) and Nickel(II) bromide trihydrate complexes (Banerjee et al., 2022).
  • The synthesis, structure, and magnetic properties of nickel (II) 6-coordinate species with several substituted aniline ligands: Studies the synthesis and magnetic properties of Nickel(II) bromide trihydrate complexes, emphasizing their potential applications (Meehan et al., 2021).

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service