Skip to Content
Merck
All Photos(1)

Documents

376728

Sigma-Aldrich

Ethylenebis(diphenylphosphine)

99%

Synonym(s):

1,2-Bis(diphenylphosphino)ethane, Diphos, dppe

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2PCH2CH2P(C6H5)2
CAS Number:
Molecular Weight:
398.42
Beilstein:
761261
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

137-142 °C (lit.)

functional group

phosphine

SMILES string

C(CP(c1ccccc1)c2ccccc2)P(c3ccccc3)c4ccccc4

InChI

1S/C26H24P2/c1-5-13-23(14-6-1)27(24-15-7-2-8-16-24)21-22-28(25-17-9-3-10-18-25)26-19-11-4-12-20-26/h1-20H,21-22H2

InChI key

QFMZQPDHXULLKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ligand for metal-catalyzed allylic alkylation; decarboxylation of allylic esters; 1,3-diene synthesis; cycloaddition reactions; carbonylation reactions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne Westphal et al.
Inorganic chemistry, 51(10), 5748-5763 (2012-05-03)
The molybdenum(V) complex [Mo(O)Cl(3)dppe] [dppe = 1,2-bis(diphenylphosphino)ethane] is considered as a model system for a combined study of the electronic structure using UV/vis absorption and magnetic circular dichroism (MCD) spectroscopy. In order to determine the signs and MCD C-term intensities
G D Hoke et al.
Molecular pharmacology, 39(1), 90-97 (1991-01-01)
Bis(diphenylphosphine)ethane (DPPE) and its gold coordination complexes have demonstrated antitumor activity in transplantable tumor models. This report describes the development of a P388 cell line (P388/DPPEc) that is resistant to DPPE and its analogues and the in vitro characterization of
Nusrat J Sanghamitra et al.
Journal of medicinal chemistry, 48(4), 977-985 (2005-02-18)
The cytotoxic properties of arylphosphines are regulated by metals. We have synthesized a series of copper(I) complexes of 1,2-bis(diphenylphosphino)ethane (DPPE) and tested their in vitro cytotoxicity in a human lung carcinoma cell line H460. One of the complexes [Cu(2)(DPPE)(3)(CH(3)CN)(2)](ClO(4))(2) (C1)
Gleb A Silantyev et al.
Inorganic chemistry, 52(4), 1787-1797 (2013-01-30)
The combination of variable temperature (190-297 K) IR and NMR spectroscopy studies with quantum-chemical calculations at the DFT/B3PW91 and AIM level had the aim to determine the mechanism of proton transfer to CpRuH(dppe) (1, dppe = Ph(2)P(CH(2))(2)PPh(2)) and the structures
David P Temelkoff et al.
Carbohydrate research, 341(10), 1645-1656 (2006-02-28)
Bis(diphenylphosphino)ethane (DPPE) reacts with pyranosyl azides derived from D-glucose and D-glucuronic acid in the presence of acid chlorides to yield the corresponding glycosyl amides. Reaction rates are comparable to those with triphenylphosphine, however, the byproduct phosphine oxide is easily removed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service