713058
1-Hexyl-3-methylimidazolium iodide
≥98% (HPLC)
Synonym(s):
HMIMI
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H19IN2
CAS Number:
Molecular Weight:
294.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥98% (HPLC)
form
liquid
impurities
≤0.5% water
SMILES string
[I-].CCCCCCn1cc[n+](C)c1
InChI
1S/C10H19N2.HI/c1-3-4-5-6-7-12-9-8-11(2)10-12;/h8-10H,3-7H2,1-2H3;1H/q+1;/p-1
InChI key
CZIUVCSYOGFUPH-UHFFFAOYSA-M
General description
1-Hexyl-3-methylimidazolium iodide is a room temperature ionic liquid (RTIL) that can be prepared by reacting 1-methylimidazole with 1-iodohexane.
Application
The addition of HMImI to the electrolyte used in dye-sensitized nanocrystalline TiO2 solar cells increases its open-circuit photovoltage.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Anomaly of charge transport of an iodide/tri-iodide redox couple in an ionic liquid and its importance in dye-sensitized solar cells
Kawano, Ryuji, and Masayoshi Watanabe
Chemical Communications (Cambridge, England), 16, 2107-2109 (2005)
The application of room temperature molten salt with low viscosity to the electrolyte for dye-sensitized solar cell
Matsumoto, Hajime, et al.
Chemistry Letters (Jpn), 30.1, 26-27 (2001)
Adsorption, thermodynamic and quantum chemical studies of 1-hexyl-3-methylimidazolium based ionic liquids as corrosion inhibitors for mild steel in HCl
Mashuga, Motsie E., et al.
Materials, 8.6, 3607-3632 (2015)
Quasi-solid-state dye-sensitized solar cells using room temperature molten salts and a low molecular weight gelator
Kubo, Wataru, et al.
Chemical Communications (Cambridge, England), 4, 374-375 (2002)
Investigation of mechanisms of enhanced open-circuit photovoltage of dye-sensitized solar cells based the electrolyte containing 1-hexyl-3-methylimidazolium iodide.
Miao W, et al.
Chinese Physics Letters, 23(3), 724-724 (2006)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service