Skip to Content
Merck
All Photos(3)

Key Documents

18122

Sigma-Aldrich

1-Butyl-3-methylimidazolium hexafluorophosphate

greener alternative

for catalysis, ≥98.5% (T)

Synonym(s):

BMIMPF6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15F6N2P
CAS Number:
Molecular Weight:
284.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.5% (T)

greener alternative product characteristics

Safer Solvents and Auxiliaries
Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤0.03% water

refractive index

n20/D 1.41

density

1.38 g/mL at 20 °C (lit.)

anion traces

bromide (Br-): ≤10 mg/kg
chloride (Cl-): ≤10 mg/kg
nitrate (NO3-): ≤10 mg/kg
phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤10 mg/kg

greener alternative category

SMILES string

F[P-](F)(F)(F)(F)F.CCCCn1cc[n+](C)c1

InChI

1S/C8H15N2.F6P/c1-3-4-5-10-7-6-9(2)8-10;1-7(2,3,4,5)6/h6-8H,3-5H2,1-2H3;/q+1;-1

InChI key

IXQYBUDWDLYNMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Butyl-3-methylimidazolium hexafluorophosphate is a high-purity ionic liquid (HPIL) with low water and halogen content, which makes it an effective solvent for transition-metal-catalyzed reactions.[1] Some of its other advantageous properties include hydrophobicity and lack of vapor pressure.[2]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermodynamic properties of 1-butyl-3-methylimidazolium hexafluorophosphate in the ideal gas state
Paulechka, Yauheni U., et al.
Journal of Chemical and Engineering Data, 48.3, 457-462 (2003)
Temperature-dependent microscopic solvent properties of `dry?and `wet?1-butyl-3-methylimidazolium hexafluorophosphate: correlation with ET (30) and Kamlet?Taft polarity scales
Baker, Sheila N., Gary A. Baker, and Frank V. Bright
Green Chemistry, 4.2, 165-169 (2002)
Olefin Metathesis in the Ionic Liquid 1?Butyl?3?methylimidazolium Hexafluorophosphate Using a Recyclable Ru Catalyst: Remarkable Effect of a Designer Ionic Tag
Yao, Qingwei, and Yiliang Zhang
Angewandte Chemie (International Edition in English), 42.29, 3395-3398 (2003)
The Negishi coupling: an update
Aldrichimica Acta, 38(3), 69-69 (2005)
Solvent properties of the 1-butyl-3-methylimidazolium hexafluorophosphate ionic liquid
Carda?Broch, S., A. Berthod, and D. W. Armstrong
Analytical and Bioanalytical Chemistry, 375.2, 191-199 (2003)

Questions

  1. Is it liquid?

    1 answer
    1. This product is supplied as either a liquid or viscous liquid.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service