Skip to Content
Merck
All Photos(1)

Documents

704326

Sigma-Aldrich

[6.6] Diphenyl C62 bis(butyric acid methyl ester)(mixture of isomers)

99.5%

Synonym(s):

Bis(1-[3-(methoxycarbonyl)propyl]-1-phenyl)-[6.6]C62 (mixture of isomers), Bis[60]PCBM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C84H28O4
Molecular Weight:
1101.12
UNSPSC Code:
12352103
NACRES:
NA.23

Quality Level

Assay

99.5%

form

solid

mp

>300 °C

solubility

organic solvents: soluble

SMILES string

COC(=O)CCCC2(c1ccccc1)C34c5c6c7c8c9c%10c(c%11c%12c3c%13c5c%14c%15c6c%16c7c%17c9c%18c%19c%20c%21c%22c%23c%24c%25c%26c%27c%28c%29c(c%20c%30c%18c%10c%31c%11c%32c%12c%33c%13c(c%26c%33c%28c%32c%29c%30%31)c%14c%25c%15c%23c%16c%22c%17%19)C%21%34C(CCCC(=O)OC)(c%35ccccc%35)C%24%27%34)C248

General description

Bis[60]PCBM is a fullerene derivative.
[6.6] Diphenyl C62bis(butyric acid methyl ester)(mixture of isomers) (bis-[60]PCBM) is a fullerene based acceptor molecule that is a bis analog of PCBM. It can be used in polymeric solar cells to improve the open circuit voltage (Voc).

Application

Soluble n-channel organic semiconductor. Used in polymer:fullerene bulk heterojunction solar cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Isomer-Pure Bis-PCBM-Assisted Crystal Engineering of Perovskite Solar Cells Showing Excellent Efficiency and Stability
Zhang F, et al.
Advanced Materials, 29(17), 1606806-1606806 (2017)
M. Lenes, G.AH. Weltzelaer, F.B. Kooistra, S.C. Veenstra, J.C. Hummelen, P.W.M. Blom
Advanced Mat., 20, 2116-2116 (2008)
Purification and electronic characterisation of 18 isomers of the OPV acceptor material bis-[60] PCBM
Shi W, et al.
Chemical Communications (Cambridge, England), 53(5), 975-978 (2017)
Material solubility effects in bulk heterojunction solar cells based on the bis-cyclopropane fullerene adducts and P3HT
Susarova DK, et al.
Solar Energy Materials and Solar Cells, 120(17), 30-36 (2014)

Articles

Progress in bulk heterojunction photodiodes since 1995 has improved device performance and widened research scope.

Progress in bulk heterojunction photodiodes since 1995 has improved device performance and widened research scope.

Progress in bulk heterojunction photodiodes since 1995 has improved device performance and widened research scope.

Progress in bulk heterojunction photodiodes since 1995 has improved device performance and widened research scope.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service