Skip to Content
Merck
All Photos(1)

Key Documents

703214

Sigma-Aldrich

Di-tetrabutylammonium cis-bis(isothiocyanato)bis(2,2′-bipyridyl-4,4′-dicarboxylato)ruthenium(II)

95% (NMR)

Synonym(s):

Greatcell Solar®, N-719 dye

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C58H86N8O8RuS2
CAS Number:
Molecular Weight:
1188.55
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

95% (NMR)

form

powder

composition

Dye content, ≥90% HPLC

mp

250-260 °C

λmax

534, 393, 313 nm (lit.)

SMILES string

S=C=N[Ru]N=C=S.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.OC(=O)c1ccnc(c1)-c2cc(ccn2)C([O-])=O.OC(=O)c3ccnc(c3)-c4cc(ccn4)C([O-])=O

InChI

1S/2C16H36N.2C12H8N2O4.2CNS.Ru/c2*1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;2*15-11(16)7-1-3-13-9(5-7)10-6-8(12(17)18)2-4-14-10;2*2-1-3;/h2*5-16H2,1-4H3;2*1-6H,(H,15,16)(H,17,18);;;/q2*+1;;;2*-1;+2/p-2

InChI key

MQGCPZMVNHGIPF-UHFFFAOYSA-L

General description

Di-tetrabutylammonium cis-bis(isothiocyanato)bis(2,2′-bipyridyl-4,4′-dicarboxylato)ruthenium(II) (N-719 dye) is a ruthenium based polypyridyl mixture used as a photosensitizer. It acts as a medium for effective absorption of photons and transfer of electrons within the electrolytic solar cells.

Application

N-719 dye is widely used as a sensitizer that can be coated on indium tin oxide (ITO) substrates by doctor blade technique, with different photoanode materials like titanium oxide (TiO2), zinc oxide (ZnO), and gold nanourchin for dye sensitized solar cell (DSSC) applications.

Legal Information

Product of Greatcell Solar Materials Pty Ltd.
Greatcell Solar is a registered trademark of Greatcell Solar Materials Pty Ltd.
Greatcell Solar is a registered trademark of Greatcell Solar

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient dye-sensitized solar cells from mesoporous zinc oxide nanostructures sensitized by N719 dye
Kumara GRA, et al.
Journal of Semiconductors, 39(3), 033005-033005 (2018)
Nanoforest of hydrothermally grown hierarchical ZnO nanowires for a high efficiency dye-sensitized solar cell
Ko SH, et al.
Nano Letters, 11(2), 666-671 (2011)
Au nanoparticle-decorated urchin-like TiO2 hierarchical microspheres for high performance dye-sensitized solar cells
Li Y, et al.
Electrochimica Acta, 293(2), 230-239 (2019)
Enhancing near-infrared solar cell response using upconverting transparentceramics
Liu M, et al.
Solar Energy Materials and Solar Cells, 95(2), 800-803 (2011)
Ruthenium sensitizers and their applications in dye-sensitized solar cells
Hara K and Koumura N
International Journal of Photoenergy (2012)

Articles

Dye-sensitized solar cells directly convert sunlight to electricity

Dye-sensitized solar cells (DSSCs) attract attention for high performance and potential low-cost production in solar energy.

Dye-sensitized solar cells (DSSCs) attract attention for high performance and potential low-cost production in solar energy.

Dye-sensitized solar cells (DSSCs) attract attention for high performance and potential low-cost production in solar energy.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service