Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Indium tin oxide coated glass slide, square
Synonym(s):
ITO, ITO coated slide, square
Linear Formula:
In
2
O
3
· (SnO
2
)
x
CAS Number:
50926-11-9
Products
Product Category
slides (3)
Length (mm)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
703192
Indium tin oxide coated glass slide, square,
surface resistivity 8-12 Ω/sq
Expand
View Pricing
703176
Indium tin oxide coated glass slide, square,
surface resistivity 70-100 Ω/sq
Expand
View Pricing
703184
Indium tin oxide coated glass slide, square,
surface resistivity 30-60 Ω/sq, slide
Expand
View Pricing