Skip to Content
Merck
All Photos(2)

Documents

692824

Sigma-Aldrich

Tantalum

powder, 60-100 mesh, 99.9%

Synonym(s):

Ta

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 537.2 °C)

Assay

99.9%

form

powder

autoignition temp.

572 °F

reaction suitability

reagent type: catalyst
core: tantalum

resistivity

13.5 μΩ-cm, 20°C

particle size

60-100 mesh

bp

5425 °C (lit.)

mp

2996 °C (lit.)

density

16.69 g/cm3 (lit.)

SMILES string

[Ta]

InChI

1S/Ta

InChI key

GUVRBAGPIYLISA-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Ashmore et al.
Journal of biomechanics, 45(16), 2931-2934 (2012-10-13)
Interest in soft tissue RadioStereometric Analysis (RSA) is rising. Previous studies have attempted to use this technique to analyse anterior cruciate ligament (ACL) graft constructs, and more recently, the movement of soft tissue as a precursor to RSA follow up
Biomimetic oxygen activation by MoS2/Ta3N5 nanocomposites for selective aerobic oxidation.
Qingsheng Gao et al.
Angewandte Chemie (International ed. in English), 51(47), 11740-11744 (2012-10-17)
S Nag et al.
Journal of the mechanical behavior of biomedical materials, 16, 21-28 (2012-11-10)
Microstructure and mechanical properties of laser deposited complex quaternary Ti-34Nb-7Zr-7Ta (all wt%), an orthopedic load-bearing implant alloy, has been investigated in detail in both as-deposited as well as heat-treated (β-solutionized and quenched) conditions. The difference in stress-strain behavior of this
Thomas Larzon et al.
Vascular, 21(2), 87-91 (2013-03-20)
Control of back bleeding from the hypogastric artery into the aneurysm after endovascular aneurysm repair (EVAR) of a ruptured aorto-iliac aneurysm may be necessary in order to avoid a type II endoleak. It is an emergency situation and selective catheterization
Hany A El-Sayed et al.
Nanoscale, 5(4), 1494-1498 (2013-01-23)
Ta oxide nanotubes (NTs) were formed by the anodization of Ta at 15 V in a solution of concentrated sulfuric acid containing 0.8-1.0 M hydrofluoric acid. To study the initial stages of NT formation, FESEM images of samples anodized for

Articles

Biomedical implants are essentially foreign substances within the human body that must survive many years’ exposure to demanding mechanical and physiological conditions. Despite these challenges, metal implants have been widely used to substitute for or rebuild hard tissues such as bones and teeth.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service