Skip to Content
Merck
All Photos(1)

Key Documents

545007

Sigma-Aldrich

Tantalum

powder, 99.99% trace metals basis

Synonym(s):

Ta

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 537.2 °C)

Quality Level

Assay

99.99% trace metals basis

form

powder

autoignition temp.

572 °F

resistivity

13.5 μΩ-cm, 20°C

bp

5425 °C (lit.)

mp

2996 °C (lit.)

density

16.69 g/cm3 (lit.)

SMILES string

[Ta]

InChI

1S/Ta

InChI key

GUVRBAGPIYLISA-UHFFFAOYSA-N

Application

Tantalum powder, a semiconducting material, is used as a photocatalyst and shows high efficiency in the solar splitting of water. Its biocompatibility makes it useful in the development of medical devices and implants.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Additively manufactured porous tantalum implants
Wauthle R, et al.
Acta Biomaterialia, 14(13), 217-225 (2015)
Tantalum-based semiconductors for solar water splitting
Zhang P, et al.
Chemical Society Reviews, 43(13), 4395-4422 (2014)
Yongyuan Guo et al.
International journal of molecular medicine, 31(3), 689-697 (2013-01-23)
Titanium (Ti) alloys of the β-type are highly attractive metallic materials for biomedical applications due to their low elastic modulus, high corrosion resistance and notable biocompatibility. A new β-type Ti35Nb2Ta3Zr alloy with a low Young's modulus of approximately 48 GPa was previously
In response.
Mariano Fernandez Fairen et al.
Spine, 38(4), 370-371 (2012-12-04)
Sining Yun et al.
ChemSusChem, 6(3), 411-416 (2013-01-31)
Ta-based compounds show Pt-like behavior: Binary tantalum compounds as counter electrodes (CEs) in dye-sensitized solar cells (DSCs) demonstrate Pt-like electrocatalytic activity and competitive photovoltaic performance, matching the performance of DSCs with Pt CEs. The first-principle density functional theory (DFT) calculations

Articles

Mechanical alloying is a “brute force” method of affecting alloying and chemical reactions. The mixture of reactant powders and several balls are placed in the milling jar of a high-energy ball mill, for example, a shaker mill or a planetary mill.

Biomedical implants are essentially foreign substances within the human body that must survive many years’ exposure to demanding mechanical and physiological conditions. Despite these challenges, metal implants have been widely used to substitute for or rebuild hard tissues such as bones and teeth.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service