Skip to Content
Merck
All Photos(1)

Key Documents

689475

Sigma-Aldrich

Methoxypolyethylene glycol azide

Synonym(s):

Polyethylene glycol, α-Methoxy-ω-azido-PEG, O-(2-Azidoethyl)-O′-methylpolyethylene glycol, PEG-Azide, mPEG Azide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
N3CH2CH2(OCH2CH2)nOCH3
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.22

description

5000

Quality Level

form

powder

mol wt

PEG average Mn 5,000

reaction suitability

reaction type: click chemistry

mp

55-60 °C

Ω-end

azide

α-end

methoxy

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service