Skip to Content
Merck
All Photos(2)

Key Documents

688509

Sigma-Aldrich

Silicon tetrachloride

packaged for use in deposition systems

Synonym(s):

STC, Tetrachlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiCl4
CAS Number:
Molecular Weight:
169.90
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor density

5.86 (vs air)

Quality Level

vapor pressure

420 mmHg ( 37.7 °C)

Assay

99.998% trace metals basis

form

liquid

reaction suitability

core: silicon

bp

57.6 °C (lit.)

mp

−70 °C (lit.)

density

1.483 g/mL at 25 °C (lit.)

SMILES string

Cl[Si](Cl)(Cl)Cl

InChI

1S/Cl4Si/c1-5(2,3)4

InChI key

FDNAPBUWERUEDA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 14 Silicon

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Kapias et al.
Journal of hazardous materials, 81(3), 209-222 (2001-02-13)
Silicon tetrachloride is a toxic, corrosive water reactive substance that is used widely in the process industries. On spillage from containment it creates liquid pools that can either boil or evaporate. The main feature of the pool behaviour is the
Claudio Curti et al.
Organic letters, 13(17), 4738-4741 (2011-08-13)
Virtually perfect transmittal of the enolate reactivity up to five conjugated double bonds from the origin allows a series of furan-based silyloxypolyenes to add to aldehyde carbonyls at the most distant point of the molecule. Denmark's axially chiral bisphosphoramide/SiCl(4) combination
Sang Ho Lee et al.
Journal of the American Chemical Society, 127(25), 9071-9078 (2005-06-23)
A series of four asymmetrically aryl-substituted 9,9'-spiro-9-silabifluorene (SSF) derivatives, 2,2'-di-tert-butyl-7,7'-diphenyl-9,9'-spiro-9-silabifluorene (PhSSF), 2,2'-di-tert-butyl-7,7'-dipyridin-2-yl-9,9'-spiro-9-silabifluorene (PySSF), 2,2'-di-tert-butyl-7,7'-dibiphenyl-4-yl-9,9'-spiro-9-silabifluorene (BPhSSF), and 2,2'-di-tert-butyl-7,7'-bis(2',2' '-bipyridin-6-yl)-9,9'-spiro-9-silabifluorene (BPySSF) are prepared through the cyclization of the corresponding 2,2'-dilithiobiphenyls with silicon tetrachloride. These novel spiro-linked silacyclopentadienes (siloles) form transparent and stable
Roman A Sandler et al.
Journal of vision, 15(9), 16-16 (2015-08-01)
Receptive field identification is a vital problem in sensory neurophysiology and vision. Much research has been done in identifying the receptive fields of nonlinear neurons whose firing rate is determined by the nonlinear interactions of a small number of linear
Sherif Zein El Abedin et al.
Accounts of chemical research, 40(11), 1106-1113 (2007-05-25)
Due to their wide thermal windows, ionic liquids can be regarded as the missing link between aqueous/organic solutions and high-temperature molten salts. They can be employed efficiently for the coating of other metals with thin layers of tantalum, aluminum, and

Articles

Atomic Layer Deposition (ALD) technology ensures uniform coating on complex 3D surfaces with precise chemisorption cycles.

Atomic Layer Deposition (ALD) technology ensures uniform coating on complex 3D surfaces with precise chemisorption cycles.

Atomic Layer Deposition (ALD) technology ensures uniform coating on complex 3D surfaces with precise chemisorption cycles.

Atomic Layer Deposition (ALD) technology ensures uniform coating on complex 3D surfaces with precise chemisorption cycles.

See All

Questions

  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service