Skip to Content

We are planning system maintenance on January 25, 2025, from 1:00 PM to 4:00 PM CST. This will impact both web and offline transactions, including online orders, quotes, price and availability checks, and order status inquiries. We apologize for any inconvenience.

Merck

683523

Sigma-Aldrich

3,5-Dimethyl-4-iodo-1H-pyrazole

97%

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H7IN2
CAS Number:
Molecular Weight:
222.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form:
solid
Assay:
97%

Quality Level

Assay

97%

form

solid

mp

136-140 °C

functional group

iodo

SMILES string

Cc1n[nH]c(C)c1I

InChI

1S/C5H7IN2/c1-3-5(6)4(2)8-7-3/h1-2H3,(H,7,8)

InChI key

MZZXIXHKDJNBJQ-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service