All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H8N4S
CAS Number:
Molecular Weight:
192.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
solid
mp
195 °C (dec.) (lit.)
211-215 °C (dec.)
functional group
phenyl
SMILES string
Nn1c(S)nnc1-c2ccccc2
InChI
1S/C8H8N4S/c9-12-7(10-11-8(12)13)6-4-2-1-3-5-6/h1-5H,9H2,(H,11,13)
InChI key
OKNHZPGPLNUEPC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Muthaiah Shellaiah et al.
Nanomaterials (Basel, Switzerland), 9(3) (2019-03-14)
Novel Cd2+ ions mediated reproducible hybrid graphite-diamond nanowire (G-DNWs; Cd2+-NDS1 NW) growth from 4-Amino-5-phenyl-4H-1,2,4-triazole-3-thiol (S1) functionalized diamond nanoparticles (NDS1) via supramolecular assembly is reported and demonstrated through TEM and AFM images. FTIR, EDX and XPS studies reveal the supramolecular coordination
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service